Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017693.hg.1 |
ENST00000391860.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017693.hg.1 |
ENST00000391860.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017693.hg.1 |
ENST00000391860.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017693.hg.1 |
ENST00000391860.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017693.hg.1 |
ENST00000391860.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017693.hg.1 |
ENST00000391860.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017693.hg.1 |
ENST00000391860.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017693.hg.1 |
ENST00000391860.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017693.hg.1 |
ENST00000391860.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017693.hg.1 |
ENST00000391860.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017693.hg.1 |
ENST00000391860.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017693.hg.1 |
ENST00000391860.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017693.hg.1 |
ENST00000525115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017693.hg.1 |
ENST00000525115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017693.hg.1 |
ENST00000525115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017693.hg.1 |
ENST00000525115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017693.hg.1 |
ENST00000525115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017693.hg.1 |
ENST00000525115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017693.hg.1 |
ENST00000525115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017693.hg.1 |
ENST00000525115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017693.hg.1 |
ENST00000530424.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017693.hg.1 |
ENST00000530424.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017693.hg.1 |
ENST00000530424.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017693.hg.1 |
ENST00000530424.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017693.hg.1 |
NM_001258311.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017693.hg.1 |
NM_001258311.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017693.hg.1 |
NM_001258311.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017693.hg.1 |
NM_001258311.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017693.hg.1 |
NM_001258311.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017693.hg.1 |
NM_001258311.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017693.hg.1 |
NM_001258311.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017693.hg.1 |
NM_001258311.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017693.hg.1 |
NM_001258311.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017693.hg.1 |
NM_001258311.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100233504.hg.1 |
ENST00000391860.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233504.hg.1 |
ENST00000525115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233504.hg.1 |
ENST00000530424.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233504.hg.1 |
NM_001258311.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100233504.hg.1 |
ENST00000391860.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233504.hg.1 |
ENST00000525115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233504.hg.1 |
ENST00000530424.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233504.hg.1 |
NM_001258311.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100233504.hg.1 |
TC0100017693.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100017693.hg.1 |
ENST00000391860.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017693.hg.1 |
ENST00000391860.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017693.hg.1 |
ENST00000391860.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017693.hg.1 |
ENST00000391860.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017693.hg.1 |
ENST00000391860.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017693.hg.1 |
ENST00000391860.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017693.hg.1 |
ENST00000391860.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017693.hg.1 |
ENST00000391860.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017693.hg.1 |
ENST00000391860.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017693.hg.1 |
ENST00000391860.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017693.hg.1 |
ENST00000391860.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017693.hg.1 |
ENST00000391860.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017693.hg.1 |
ENST00000525115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017693.hg.1 |
ENST00000525115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017693.hg.1 |
ENST00000525115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017693.hg.1 |
ENST00000525115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017693.hg.1 |
ENST00000525115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017693.hg.1 |
ENST00000525115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017693.hg.1 |
ENST00000525115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017693.hg.1 |
ENST00000525115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017693.hg.1 |
ENST00000530424.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017693.hg.1 |
ENST00000530424.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017693.hg.1 |
ENST00000530424.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017693.hg.1 |
ENST00000530424.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017693.hg.1 |
NM_001258311.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017693.hg.1 |
NM_001258311.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017693.hg.1 |
NM_001258311.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017693.hg.1 |
NM_001258311.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017693.hg.1 |
NM_001258311.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017693.hg.1 |
NM_001258311.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017693.hg.1 |
NM_001258311.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017693.hg.1 |
NM_001258311.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017693.hg.1 |
NM_001258311.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017693.hg.1 |
NM_001258311.1 |
Refseq |