Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031837.hg.1 |
TC03001755.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031837.hg.1 |
ENST00000341105.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031837.hg.1 |
ENST00000430265.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031837.hg.1 |
ENST00000487848.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031837.hg.1 |
NM_001145661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031837.hg.1 |
NM_001145662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031837.hg.1 |
NM_032638.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031838.hg.1 |
TC03001755.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031838.hg.1 |
ENST00000341105.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031838.hg.1 |
ENST00000430265.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031838.hg.1 |
ENST00000487848.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031838.hg.1 |
NM_001145661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031838.hg.1 |
NM_001145662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031838.hg.1 |
NM_032638.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000341105.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000341105.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000341105.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000341105.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000341105.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000341105.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000341105.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000341105.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000341105.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000341105.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000341105.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000341105.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000341105.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000430265.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000430265.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000430265.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000430265.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000430265.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000430265.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000430265.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000430265.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000430265.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000430265.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000487848.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000487848.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000487848.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000487848.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000487848.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000487848.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000487848.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000487848.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000487848.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000487848.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000489987.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000489987.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000489987.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000489987.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000489987.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000489987.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000492608.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000492608.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000492608.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000492608.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000498200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000498200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000498200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
ENST00000498200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
NM_001145661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
NM_001145661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
NM_001145661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
NM_001145661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
NM_001145661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
NM_001145661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
NM_001145661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
NM_001145661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
NM_001145661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
NM_001145661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
NM_001145661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
NM_001145661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
NM_001145661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
NM_001145662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
NM_001145662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
NM_001145662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
NM_001145662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
NM_001145662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
NM_001145662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
NM_001145662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
NM_001145662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
NM_001145662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
NM_001145662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
NM_001145662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
NM_032638.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
NM_032638.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
NM_032638.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
NM_032638.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
NM_032638.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
NM_032638.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
NM_032638.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
NM_032638.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
NM_032638.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
NM_032638.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
NM_032638.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
NM_032638.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012339.hg.1 |
NM_032638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000341105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000341105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000341105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000341105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000341105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000341105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000341105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000341105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000341105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000341105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000341105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000341105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000341105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000430265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000430265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000430265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000430265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000430265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000430265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000430265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000430265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000430265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000430265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000487848.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000487848.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000487848.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000487848.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000487848.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000487848.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000487848.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000487848.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000487848.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000487848.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000489987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000489987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000489987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000489987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000489987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000489987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000492608.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000492608.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000492608.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000492608.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000498200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000498200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000498200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
ENST00000498200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
NM_001145661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
NM_001145661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
NM_001145661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
NM_001145661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
NM_001145661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
NM_001145661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
NM_001145661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
NM_001145661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
NM_001145661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
NM_001145661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
NM_001145661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
NM_001145661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
NM_001145661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
NM_001145662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
NM_001145662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
NM_001145662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
NM_001145662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
NM_001145662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
NM_001145662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
NM_001145662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
NM_001145662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
NM_001145662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
NM_001145662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
NM_001145662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
NM_032638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
NM_032638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
NM_032638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
NM_032638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
NM_032638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
NM_032638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
NM_032638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
NM_032638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
NM_032638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
NM_032638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
NM_032638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
NM_032638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012339.hg.1 |
NM_032638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300192292.hg.1 |
ENST00000341105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300192292.hg.1 |
ENST00000430265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300192292.hg.1 |
ENST00000487848.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300192292.hg.1 |
NM_001145661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300192292.hg.1 |
NM_001145662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300192292.hg.1 |
NM_032638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300192292.hg.1 |
ENST00000341105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300192292.hg.1 |
ENST00000430265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300192292.hg.1 |
ENST00000487848.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300192292.hg.1 |
NM_001145661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300192292.hg.1 |
NM_001145662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300192292.hg.1 |
NM_032638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300192292.hg.1 |
TC0300012339.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300192293.hg.1 |
ENST00000341105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300192293.hg.1 |
ENST00000430265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300192293.hg.1 |
ENST00000487848.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300192293.hg.1 |
ENST00000492608.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300192293.hg.1 |
NM_001145661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300192293.hg.1 |
NM_001145662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300192293.hg.1 |
NM_032638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300192293.hg.1 |
ENST00000341105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300192293.hg.1 |
ENST00000430265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300192293.hg.1 |
ENST00000487848.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300192293.hg.1 |
ENST00000492608.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300192293.hg.1 |
NM_001145661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300192293.hg.1 |
NM_001145662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300192293.hg.1 |
NM_032638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300192293.hg.1 |
TC0300012339.hg.1 |
Clariom D TC |