Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000359927.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000359927.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000359927.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09002557.hg.1 |
TC09000216.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09002557.hg.1 |
ENST00000377765.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09002557.hg.1 |
ENST00000536622.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09002557.hg.1 |
ENST00000539465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09002557.hg.1 |
ENST00000541302.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09002557.hg.1 |
NM_014907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900148854.hg.1 |
ENST00000377765.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900148854.hg.1 |
ENST00000539465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900148854.hg.1 |
NM_014907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900148854.hg.1 |
ENST00000377765.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900148854.hg.1 |
ENST00000539465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900148854.hg.1 |
NM_014907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900148854.hg.1 |
TC0900007163.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000359927.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000359927.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000359927.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |