Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019467.hg.1 |
TC09001547.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019467.hg.1 |
ENST00000286713.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019467.hg.1 |
ENST00000538954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019467.hg.1 |
NM_004099.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
ENST00000286713.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
ENST00000286713.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
ENST00000286713.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
ENST00000286713.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
ENST00000286713.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
ENST00000286713.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
ENST00000286713.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
ENST00000286713.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
ENST00000286713.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
ENST00000286713.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
ENST00000286713.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
ENST00000286713.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
ENST00000286713.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
ENST00000347359.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
ENST00000347359.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
ENST00000347359.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
ENST00000347359.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
ENST00000347359.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
ENST00000538954.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
ENST00000538954.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
ENST00000538954.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
ENST00000538954.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
ENST00000538954.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
ENST00000538954.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
ENST00000538954.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
ENST00000538954.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
ENST00000538954.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_001270526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_001270526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_001270526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_001270526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_001270526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_001270526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_001270526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_001270526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_001270526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_001270526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_001270526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_001270526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_001270526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_001270526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_001270526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_001270527.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_001270527.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_001270527.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_001270527.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_001270527.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_001270527.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_001270527.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_001270527.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_001270527.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_004099.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_004099.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_004099.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_004099.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_004099.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_004099.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_004099.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_004099.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_004099.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_004099.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_004099.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_004099.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_004099.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_004099.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_004099.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_004099.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_198194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_198194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_198194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_198194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_198194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_198194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_198194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_198194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_198194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NM_198194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NR_073037.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NR_073037.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NR_073037.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NR_073037.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NR_073037.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NR_073037.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NR_073037.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NR_073037.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NR_073037.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NR_073037.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NR_073037.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NR_073037.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NR_073037.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NR_073037.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NR_073037.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NR_073037.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011404.hg.1 |
NR_073037.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177795.hg.1 |
ENST00000286713.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900177795.hg.1 |
NM_001270526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177795.hg.1 |
NM_004099.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177795.hg.1 |
NR_073037.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177795.hg.1 |
ENST00000286713.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900177795.hg.1 |
NM_001270526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177795.hg.1 |
NM_004099.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177795.hg.1 |
NR_073037.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177795.hg.1 |
TC0900011404.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900177796.hg.1 |
ENST00000286713.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900177796.hg.1 |
NM_001270526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177796.hg.1 |
NM_004099.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177796.hg.1 |
NR_073037.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177796.hg.1 |
ENST00000286713.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900177796.hg.1 |
NM_001270526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177796.hg.1 |
NM_004099.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177796.hg.1 |
NR_073037.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177796.hg.1 |
TC0900011404.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
ENST00000286713.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
ENST00000286713.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
ENST00000286713.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
ENST00000286713.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
ENST00000286713.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
ENST00000286713.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
ENST00000286713.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
ENST00000286713.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
ENST00000286713.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
ENST00000286713.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
ENST00000286713.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
ENST00000286713.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
ENST00000286713.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
ENST00000347359.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
ENST00000347359.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
ENST00000347359.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
ENST00000347359.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
ENST00000347359.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
ENST00000538954.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
ENST00000538954.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
ENST00000538954.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
ENST00000538954.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
ENST00000538954.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
ENST00000538954.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
ENST00000538954.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
ENST00000538954.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
ENST00000538954.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_001270526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_001270526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_001270526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_001270526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_001270526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_001270526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_001270526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_001270526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_001270526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_001270526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_001270526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_001270526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_001270526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_001270526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_001270526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_001270527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_001270527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_001270527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_001270527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_001270527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_001270527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_001270527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_001270527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_001270527.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_004099.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_004099.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_004099.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_004099.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_004099.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_004099.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_004099.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_004099.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_004099.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_004099.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_004099.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_004099.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_004099.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_004099.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_004099.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_004099.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_198194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_198194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_198194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_198194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_198194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_198194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_198194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_198194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_198194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NM_198194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NR_073037.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NR_073037.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NR_073037.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NR_073037.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NR_073037.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NR_073037.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NR_073037.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NR_073037.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NR_073037.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NR_073037.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NR_073037.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NR_073037.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NR_073037.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NR_073037.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NR_073037.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NR_073037.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011404.hg.1 |
NR_073037.1 |
Refseq |