Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026051.hg.1 |
ENST00000305447.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026051.hg.1 |
ENST00000393297.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026051.hg.1 |
ENST00000418177.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026051.hg.1 |
NM_001143831.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026051.hg.1 |
TC11002176.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026050.hg.1 |
ENST00000305447.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026050.hg.1 |
ENST00000418177.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026050.hg.1 |
NM_001143831.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026050.hg.1 |
TC11002176.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000305432.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000305432.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000305432.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000305432.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000305432.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000305432.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000305432.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000305432.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000305432.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000305432.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000305432.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000305432.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000305432.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000305447.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000305447.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000305447.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000305447.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000305447.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000305447.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000305447.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000305447.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000305447.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000305447.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000305447.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000305447.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000305447.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000305447.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000393294.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000393294.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000393294.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000393294.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000393294.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000393294.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000393294.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000449371.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000449371.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000449371.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000449371.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000455756.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000455756.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000455756.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000455756.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000455756.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000455756.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000455756.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000455756.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000455756.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000455756.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000455756.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000455756.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000455756.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000455756.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000455756.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
ENST00000455756.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
NM_000842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
NM_000842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
NM_000842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
NM_000842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
NM_000842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
NM_000842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
NM_000842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
NM_000842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
NM_000842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
NM_000842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
NM_000842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
NM_000842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
NM_000842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
NM_000842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
NM_000842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
NM_000842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
NM_000842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
NM_001143831.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
NM_001143831.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
NM_001143831.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
NM_001143831.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
NM_001143831.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
NM_001143831.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
NM_001143831.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
NM_001143831.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
NM_001143831.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
NM_001143831.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
NM_001143831.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
NM_001143831.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
NM_001143831.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
NM_001143831.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
NM_001143831.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011895.hg.1 |
NM_001143831.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100191847.hg.1 |
ENST00000305447.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100191847.hg.1 |
NM_001143831.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100191847.hg.1 |
ENST00000305447.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100191847.hg.1 |
NM_001143831.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100191847.hg.1 |
TC1100011895.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100191849.hg.1 |
ENST00000305432.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100191849.hg.1 |
ENST00000305447.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100191849.hg.1 |
ENST00000455756.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100191849.hg.1 |
NM_000842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100191849.hg.1 |
NM_001143831.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100191849.hg.1 |
ENST00000305432.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100191849.hg.1 |
ENST00000305447.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100191849.hg.1 |
ENST00000455756.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100191849.hg.1 |
NM_000842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100191849.hg.1 |
NM_001143831.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100191849.hg.1 |
TC1100011895.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000305432.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000305432.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000305432.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000305432.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000305432.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000305432.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000305432.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000305432.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000305432.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000305432.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000305432.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000305432.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000305432.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000305447.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000305447.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000305447.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000305447.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000305447.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000305447.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000305447.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000305447.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000305447.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000305447.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000305447.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000305447.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000305447.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000305447.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000393294.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000393294.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000393294.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000393294.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000393294.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000393294.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000393294.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000449371.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000449371.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000449371.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000449371.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000455756.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000455756.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000455756.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000455756.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000455756.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000455756.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000455756.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000455756.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000455756.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000455756.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000455756.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000455756.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000455756.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000455756.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000455756.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
ENST00000455756.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
NM_000842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
NM_000842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
NM_000842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
NM_000842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
NM_000842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
NM_000842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
NM_000842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
NM_000842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
NM_000842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
NM_000842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
NM_000842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
NM_000842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
NM_000842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
NM_000842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
NM_000842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
NM_000842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
NM_000842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
NM_001143831.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
NM_001143831.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
NM_001143831.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
NM_001143831.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
NM_001143831.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
NM_001143831.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
NM_001143831.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
NM_001143831.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
NM_001143831.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
NM_001143831.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
NM_001143831.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
NM_001143831.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
NM_001143831.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
NM_001143831.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
NM_001143831.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011895.hg.1 |
NM_001143831.2 |
Refseq |