Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032737.3 | NP_116126.3 | 4 - 5 | 724 | 117 | |
GenBank mRNA | AL560944.3 | - | 723 | 117 | ||
BC006551.2 | - | 4 - 5 | 689 | 2961134 | 117 | |
BT007441.1 | - | 4 - 5 | 629 | 117 | ||
M94362.1 | - | 3 - 4 | 377 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |