Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRPF40A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017892.3 | NP_060362.3 | 4 - 5 | 352 | 129 | |
XM_005246676.4 | XP_005246733.3 | 4 - 5 | 872 | 129 | ||
XM_005246677.4 | XP_005246734.2 | 4 - 5 | 348 | 129 | ||
XM_011511446.2 | XP_011509748.1 | 4 - 5 | 872 | 129 | ||
XM_011511447.2 | XP_011509749.1 | 4 - 5 | 872 | 129 | ||
XM_011511448.2 | XP_011509750.1 | 4 - 5 | 348 | 129 | ||
XM_011511449.2 | XP_011509751.1 | 6 - 7 | 702 | 129 | ||
XM_017004453.1 | XP_016859942.1 | 4 - 5 | 872 | 129 | ||
XM_017004454.1 | XP_016859943.1 | 4 - 5 | 872 | 129 | ||
XM_017004455.1 | XP_016859944.1 | 4 - 5 | 348 | 129 | ||
XM_017004456.1 | XP_016859945.1 | 5 - 6 | 538 | 129 | ||
XM_017004457.1 | XP_016859946.1 | 6 - 7 | 701 | 129 | ||
XM_017004458.1 | XP_016859947.1 | 5 - 6 | 502 | 129 | ||
XM_017004459.1 | XP_016859948.1 | 5 - 6 | 611 | 129 | ||
XM_017004460.1 | XP_016859949.1 | 5 - 6 | 539 | 129 | ||
XM_017004461.1 | XP_016859950.1 | 5 - 6 | 611 | 129 | ||
GenBank mRNA | AF049523.1 | - | 4 - 5 | 337 | 129 | |
AF049524.1 | - | 4 - 5 | 290 | 129 | ||
AF049528.1 | - | 4 - 5 | 424 | 129 | ||
BC000273.1 | - | 4 - 5 | 315 | 129 | ||
BC011788.2 | - | 4 - 5 | 336 | 129 | ||
BC029414.1 | - | 4 - 5 | 352 | 129 | ||
U70667.1 | - | 4 - 5 | 390 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |