Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016094.hg.1 |
TC08001245.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016094.hg.1 |
ENST00000301645.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016094.hg.1 |
NM_000780.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016095.hg.1 |
TC08001245.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016095.hg.1 |
ENST00000301645.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016095.hg.1 |
NM_000780.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010551.hg.1 |
ENST00000301645.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010551.hg.1 |
ENST00000301645.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010551.hg.1 |
ENST00000301645.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010551.hg.1 |
ENST00000301645.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010551.hg.1 |
ENST00000301645.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010551.hg.1 |
ENST00000301645.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010551.hg.1 |
ENST00000301645.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010551.hg.1 |
ENST00000301645.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010551.hg.1 |
ENST00000301645.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010551.hg.1 |
ENST00000301645.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010551.hg.1 |
NM_000780.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010551.hg.1 |
NM_000780.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010551.hg.1 |
NM_000780.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010551.hg.1 |
NM_000780.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010551.hg.1 |
NM_000780.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010551.hg.1 |
NM_000780.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010551.hg.1 |
NM_000780.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010551.hg.1 |
NM_000780.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010551.hg.1 |
NM_000780.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800171673.hg.1 |
ENST00000301645.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800171673.hg.1 |
NM_000780.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800171673.hg.1 |
ENST00000301645.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800171673.hg.1 |
NM_000780.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800171673.hg.1 |
TC0800010551.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800010551.hg.1 |
ENST00000301645.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010551.hg.1 |
ENST00000301645.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010551.hg.1 |
ENST00000301645.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010551.hg.1 |
ENST00000301645.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010551.hg.1 |
ENST00000301645.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010551.hg.1 |
ENST00000301645.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010551.hg.1 |
ENST00000301645.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010551.hg.1 |
ENST00000301645.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010551.hg.1 |
ENST00000301645.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010551.hg.1 |
ENST00000301645.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010551.hg.1 |
NM_000780.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010551.hg.1 |
NM_000780.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010551.hg.1 |
NM_000780.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010551.hg.1 |
NM_000780.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010551.hg.1 |
NM_000780.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010551.hg.1 |
NM_000780.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010551.hg.1 |
NM_000780.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010551.hg.1 |
NM_000780.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010551.hg.1 |
NM_000780.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800171672.hg.1 |
ENST00000301645.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800171672.hg.1 |
NM_000780.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800171672.hg.1 |
ENST00000301645.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800171672.hg.1 |
NM_000780.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800171672.hg.1 |
TC0800010551.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800171674.hg.1 |
ENST00000301645.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800171674.hg.1 |
NM_000780.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800171674.hg.1 |
ENST00000301645.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800171674.hg.1 |
NM_000780.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800171674.hg.1 |
TC0800010551.hg.1 |
Clariom D TC |