Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
ENST00000367942.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
ENST00000367942.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
ENST00000367942.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
ENST00000367942.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
ENST00000367942.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
ENST00000367942.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
ENST00000367942.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
ENST00000367942.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
ENST00000367942.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
ENST00000367942.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
ENST00000367942.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
ENST00000367942.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
ENST00000367942.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
ENST00000367942.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
ENST00000367942.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
ENST00000367942.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
ENST00000367942.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
ENST00000367942.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
ENST00000367942.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
ENST00000367942.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
ENST00000367942.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
ENST00000476437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
ENST00000476437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
ENST00000476437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
ENST00000476437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
ENST00000476437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
NM_007348.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
NM_007348.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
NM_007348.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
NM_007348.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
NM_007348.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
NM_007348.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
NM_007348.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
NM_007348.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
NM_007348.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
NM_007348.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
NM_007348.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
NM_007348.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
NM_007348.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
NM_007348.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
NM_007348.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
NM_007348.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
NM_007348.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
NM_007348.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
NM_007348.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
NM_007348.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
NM_007348.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010369.hg.1 |
NM_007348.3 |
Refseq |