Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013130.hg.1 |
TC06001110.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013130.hg.1 |
ENST00000367243.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013130.hg.1 |
ENST00000367244.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013130.hg.1 |
NM_003381.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013130.hg.1 |
NM_194435.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
ENST00000367243.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
ENST00000367243.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
ENST00000367243.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
ENST00000367243.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
ENST00000367243.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
ENST00000367243.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
ENST00000367243.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
ENST00000367243.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
ENST00000367243.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
ENST00000367243.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
ENST00000367244.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
ENST00000367244.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
ENST00000367244.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
ENST00000367244.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
ENST00000367244.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
ENST00000367244.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
ENST00000367244.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
ENST00000367244.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
ENST00000367244.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
ENST00000367244.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
ENST00000431366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
ENST00000431366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
ENST00000431366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
ENST00000431366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
ENST00000431366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
ENST00000431366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
ENST00000431366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
NM_003381.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
NM_003381.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
NM_003381.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
NM_003381.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
NM_003381.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
NM_003381.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
NM_003381.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
NM_003381.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
NM_003381.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
NM_003381.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
NM_194435.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
NM_194435.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
NM_194435.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
NM_194435.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
NM_194435.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
NM_194435.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
NM_194435.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
NM_194435.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
NM_194435.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009886.hg.1 |
NM_194435.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600167654.hg.1 |
ENST00000367243.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600167654.hg.1 |
ENST00000367244.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600167654.hg.1 |
ENST00000431366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600167654.hg.1 |
NM_003381.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600167654.hg.1 |
NM_194435.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600167654.hg.1 |
ENST00000367243.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600167654.hg.1 |
ENST00000367244.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600167654.hg.1 |
ENST00000431366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600167654.hg.1 |
NM_003381.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600167654.hg.1 |
NM_194435.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600167654.hg.1 |
TC0600009886.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
ENST00000367243.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
ENST00000367243.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
ENST00000367243.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
ENST00000367243.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
ENST00000367243.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
ENST00000367243.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
ENST00000367243.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
ENST00000367243.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
ENST00000367243.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
ENST00000367243.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
ENST00000367244.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
ENST00000367244.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
ENST00000367244.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
ENST00000367244.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
ENST00000367244.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
ENST00000367244.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
ENST00000367244.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
ENST00000367244.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
ENST00000367244.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
ENST00000367244.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
ENST00000431366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
ENST00000431366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
ENST00000431366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
ENST00000431366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
ENST00000431366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
ENST00000431366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
ENST00000431366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
NM_003381.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
NM_003381.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
NM_003381.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
NM_003381.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
NM_003381.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
NM_003381.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
NM_003381.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
NM_003381.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
NM_003381.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
NM_003381.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
NM_194435.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
NM_194435.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
NM_194435.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
NM_194435.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
NM_194435.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
NM_194435.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
NM_194435.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
NM_194435.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
NM_194435.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009886.hg.1 |
NM_194435.2 |
Refseq |