Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005268208.1 | XP_005268265.1 | 13 - 14 | 1749 | 89 | |
XM_005268209.3 | XP_005268266.1 | 13 - 14 | 1553 | 89 | ||
XM_017021765.1 | XP_016877254.1 | 13 - 14 | 1516 | 89 | ||
GenBank mRNA | AB082526.1 | - | 13 - 14 | 1685 | 89 | |
AK128693.1 | - | 13 - 14 | 2153 | 89 | ||
AL832633.2 | - | 7 - 8 | 1374 | 89 | ||
AY048580.1 | - | 13 - 14 | 1579 | 89 | ||
AY080896.1 | - | 13 - 14 | 1591 | 89 | ||
BC093881.1 | - | 13 - 14 | 1660 | 7939726 | 89 | |
BC112101.1 | - | 13 - 14 | 1660 | 8327569 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons |