Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12007710.hg.1 |
ENST00000308330.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12007710.hg.1 |
NM_001167614.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12007710.hg.1 |
NM_014319.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12007710.hg.1 |
TC12000581.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000308330.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000308330.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000308330.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000308330.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000308330.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000308330.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000308330.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000308330.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000308330.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000308330.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000308330.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000308330.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000308330.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000308330.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000308330.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000308330.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000308330.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000308330.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000539442.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000539442.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000539442.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000539442.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000539442.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000539442.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000539442.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000539442.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000539442.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000541171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000542032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000542032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000542032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000542032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000544506.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000544506.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000544506.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000544506.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000544506.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000545026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000545026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000545026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000545026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000545026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000545026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000545026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
ENST00000545026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
NM_001167614.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
NM_001167614.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
NM_001167614.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
NM_001167614.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
NM_001167614.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
NM_001167614.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
NM_001167614.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
NM_001167614.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
NM_001167614.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
NM_001167614.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
NM_001167614.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
NM_001167614.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
NM_001167614.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
NM_001167614.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
NM_001167614.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
NM_001167614.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
NM_001167614.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
NM_001167614.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
NM_001167614.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
NM_014319.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
NM_014319.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
NM_014319.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
NM_014319.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
NM_014319.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
NM_014319.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
NM_014319.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
NM_014319.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
NM_014319.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
NM_014319.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
NM_014319.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
NM_014319.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
NM_014319.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
NM_014319.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
NM_014319.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
NM_014319.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
NM_014319.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
NM_014319.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008026.hg.1 |
NM_014319.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200158798.hg.1 |
ENST00000308330.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200158798.hg.1 |
NM_001167614.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200158798.hg.1 |
NM_014319.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200158798.hg.1 |
ENST00000308330.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200158798.hg.1 |
NM_001167614.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200158798.hg.1 |
NM_014319.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200158798.hg.1 |
TC1200008026.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000308330.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000308330.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000308330.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000308330.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000308330.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000308330.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000308330.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000308330.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000308330.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000308330.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000308330.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000308330.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000308330.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000308330.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000308330.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000308330.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000308330.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000308330.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000539442.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000539442.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000539442.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000539442.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000539442.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000539442.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000539442.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000539442.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000539442.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000541171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000542032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000542032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000542032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000542032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000544506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000544506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000544506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000544506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000544506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000545026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000545026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000545026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000545026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000545026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000545026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000545026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
ENST00000545026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
NM_001167614.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
NM_001167614.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
NM_001167614.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
NM_001167614.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
NM_001167614.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
NM_001167614.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
NM_001167614.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
NM_001167614.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
NM_001167614.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
NM_001167614.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
NM_001167614.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
NM_001167614.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
NM_001167614.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
NM_001167614.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
NM_001167614.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
NM_001167614.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
NM_001167614.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
NM_001167614.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
NM_001167614.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
NM_014319.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
NM_014319.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
NM_014319.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
NM_014319.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
NM_014319.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
NM_014319.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
NM_014319.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
NM_014319.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
NM_014319.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
NM_014319.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
NM_014319.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
NM_014319.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
NM_014319.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
NM_014319.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
NM_014319.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
NM_014319.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
NM_014319.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
NM_014319.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008026.hg.1 |
NM_014319.4 |
Refseq |