Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377238.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377238.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377238.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377238.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377238.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377238.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377238.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377238.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377238.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377238.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377238.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377238.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377238.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377238.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377238.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377242.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377242.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377242.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377242.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377242.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377242.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377242.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377242.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377242.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377242.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377242.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377242.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377242.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377242.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377242.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377242.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377242.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377242.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377252.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377252.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377252.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377252.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377252.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377252.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377252.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377252.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377252.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377252.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377252.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377252.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377252.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377252.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377252.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377252.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377252.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377252.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377252.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
ENST00000377252.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
NM_001282736.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
NM_001282736.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
NM_001282736.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
NM_001282736.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
NM_001282736.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
NM_001282736.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
NM_001282736.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
NM_001282736.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
NM_001282736.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
NM_001282736.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
NM_001282736.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
NM_001282736.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
NM_001282736.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
NM_001282736.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
NM_001282736.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
NM_001282736.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
NM_001282736.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
NM_001282736.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
NM_032812.8 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
NM_032812.8 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
NM_032812.8 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
NM_032812.8 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
NM_032812.8 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
NM_032812.8 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
NM_032812.8 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
NM_032812.8 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
NM_032812.8 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
NM_032812.8 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
NM_032812.8 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
NM_032812.8 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
NM_032812.8 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
NM_032812.8 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
NM_032812.8 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
NM_032812.8 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
NM_032812.8 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
NM_032812.8 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006937.hg.1 |
NM_032812.8 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10001685.hg.1 |
ENST00000377238.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10001685.hg.1 |
ENST00000377242.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10001685.hg.1 |
ENST00000377252.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10001685.hg.1 |
ENST00000536022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10001685.hg.1 |
NM_032812.8 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10001685.hg.1 |
TC10000145.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377238.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377238.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377238.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377238.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377238.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377238.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377238.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377238.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377238.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377238.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377238.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377238.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377238.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377238.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377238.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377242.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377242.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377242.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377242.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377242.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377242.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377242.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377242.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377242.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377242.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377242.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377242.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377242.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377242.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377242.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377242.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377242.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377242.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377252.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377252.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377252.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377252.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377252.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377252.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377252.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377252.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377252.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377252.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377252.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377252.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377252.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377252.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377252.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377252.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377252.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377252.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377252.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
ENST00000377252.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
NM_001282736.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
NM_001282736.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
NM_001282736.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
NM_001282736.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
NM_001282736.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
NM_001282736.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
NM_001282736.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
NM_001282736.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
NM_001282736.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
NM_001282736.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
NM_001282736.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
NM_001282736.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
NM_001282736.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
NM_001282736.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
NM_001282736.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
NM_001282736.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
NM_001282736.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
NM_001282736.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
NM_032812.8 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
NM_032812.8 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
NM_032812.8 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
NM_032812.8 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
NM_032812.8 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
NM_032812.8 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
NM_032812.8 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
NM_032812.8 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
NM_032812.8 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
NM_032812.8 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
NM_032812.8 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
NM_032812.8 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
NM_032812.8 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
NM_032812.8 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
NM_032812.8 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
NM_032812.8 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
NM_032812.8 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
NM_032812.8 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006937.hg.1 |
NM_032812.8 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000147525.hg.1 |
ENST00000377238.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000147525.hg.1 |
ENST00000377242.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000147525.hg.1 |
ENST00000377252.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000147525.hg.1 |
NM_001282736.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000147525.hg.1 |
NM_032812.8 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000147525.hg.1 |
ENST00000377238.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000147525.hg.1 |
ENST00000377242.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000147525.hg.1 |
ENST00000377252.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000147525.hg.1 |
NM_001282736.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000147525.hg.1 |
NM_032812.8 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000147525.hg.1 |
TC1000006937.hg.1 |
Clariom D TC |