Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000528137.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000528137.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000528137.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000528137.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000528573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000528573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000528573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000528573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000528573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000528573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000529532.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000529532.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000529532.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000529532.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000529532.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000529532.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000529532.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000529532.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000529532.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000529532.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000529532.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000529532.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000529532.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000529532.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000529580.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000529580.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000529580.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000529580.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000530251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000530251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000530251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000530251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000530251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000530251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000530251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000530251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000532333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000532333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000532333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000532333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000532333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000533386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000533386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000533386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
ENST00000533386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100158625.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100158625.hg.1 |
ENST00000529532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100158625.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100158625.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100158625.hg.1 |
ENST00000529532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100158625.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100158625.hg.1 |
TC1100008046.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000312397.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000528137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000528137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000528137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000528137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000528573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000528573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000528573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000528573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000528573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000528573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000529532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000529532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000529532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000529532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000529532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000529532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000529532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000529532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000529532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000529532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000529532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000529532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000529532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000529532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000529580.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000529580.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000529580.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000529580.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000530251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000530251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000530251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000530251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000530251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000530251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000530251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000530251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000532333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000532333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000532333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000532333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000532333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000533386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000533386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000533386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
ENST00000533386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008046.hg.1 |
NM_005146.4 |
Refseq |