Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000368843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000458013.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000458013.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000458013.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000458013.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000458013.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000458013.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000458013.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000458013.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000458013.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000458013.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000458013.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000458013.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000458013.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000458013.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000458013.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000458013.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000482791.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000482791.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000482791.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
ENST00000482791.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
NM_030918.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
NM_030918.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
NM_030918.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
NM_030918.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
NM_030918.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
NM_030918.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
NM_030918.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
NM_030918.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
NM_030918.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
NM_030918.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
NM_030918.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
NM_030918.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
NM_030918.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
NM_030918.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
NM_030918.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
NM_030918.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009965.hg.1 |
NM_030918.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01018857.hg.1 |
TC01001214.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01018857.hg.1 |
ENST00000368838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01018857.hg.1 |
ENST00000368843.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01018857.hg.1 |
ENST00000458013.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01018857.hg.1 |
NM_030918.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100172274.hg.1 |
ENST00000368838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172274.hg.1 |
ENST00000368841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172274.hg.1 |
ENST00000368843.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172274.hg.1 |
ENST00000458013.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172274.hg.1 |
NM_030918.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100172274.hg.1 |
ENST00000368838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172274.hg.1 |
ENST00000368841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172274.hg.1 |
ENST00000368843.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172274.hg.1 |
ENST00000458013.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172274.hg.1 |
NM_030918.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100172274.hg.1 |
TC0100009965.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368843.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368843.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368843.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368843.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368843.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368843.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368843.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368843.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368843.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368843.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368843.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368843.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368843.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368843.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368843.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368843.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368843.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368843.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000368843.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000458013.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000458013.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000458013.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000458013.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000458013.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000458013.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000458013.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000458013.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000458013.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000458013.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000458013.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000458013.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000458013.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000458013.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000458013.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000458013.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000482791.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000482791.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000482791.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
ENST00000482791.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
NM_030918.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
NM_030918.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
NM_030918.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
NM_030918.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
NM_030918.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
NM_030918.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
NM_030918.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
NM_030918.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
NM_030918.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
NM_030918.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
NM_030918.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
NM_030918.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
NM_030918.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
NM_030918.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
NM_030918.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
NM_030918.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009965.hg.1 |
NM_030918.5 |
Refseq |