Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002681.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002681.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002681.hg.1 |
TC11000199.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002682.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002682.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002682.hg.1 |
TC11000199.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002684.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002684.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002684.hg.1 |
TC11000199.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006876.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006876.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006876.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006876.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006876.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006876.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006876.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006876.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006876.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006876.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006876.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006876.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006876.hg.1 |
ENST00000529028.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006876.hg.1 |
ENST00000529028.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006876.hg.1 |
ENST00000529028.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006876.hg.1 |
ENST00000529028.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006876.hg.1 |
ENST00000533534.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006876.hg.1 |
ENST00000533534.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006876.hg.1 |
ENST00000533534.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006876.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006876.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006876.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006876.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006876.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006876.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006876.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006876.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006876.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006876.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006876.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006876.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002678.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002678.hg.1 |
ENST00000529028.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002678.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002678.hg.1 |
TC11000199.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002677.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002677.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002677.hg.1 |
TC11000199.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002683.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002683.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002683.hg.1 |
TC11000199.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100148414.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100148414.hg.1 |
ENST00000529028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100148414.hg.1 |
ENST00000533534.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100148414.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100148414.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100148414.hg.1 |
ENST00000529028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100148414.hg.1 |
ENST00000533534.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100148414.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100148414.hg.1 |
TC1100006876.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100148421.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100148421.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100148421.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100148421.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100148421.hg.1 |
TC1100006876.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100148418.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100148418.hg.1 |
ENST00000533534.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100148418.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100148418.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100148418.hg.1 |
ENST00000533534.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100148418.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100148418.hg.1 |
TC1100006876.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100148419.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100148419.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100148419.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100148419.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100148419.hg.1 |
TC1100006876.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100148412.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100148412.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100148412.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100148412.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100148412.hg.1 |
TC1100006876.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100148420.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100148420.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100148420.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100148420.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100148420.hg.1 |
TC1100006876.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100006876.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006876.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006876.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006876.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006876.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006876.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006876.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006876.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006876.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006876.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006876.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006876.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006876.hg.1 |
ENST00000529028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006876.hg.1 |
ENST00000529028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006876.hg.1 |
ENST00000529028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006876.hg.1 |
ENST00000529028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006876.hg.1 |
ENST00000533534.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006876.hg.1 |
ENST00000533534.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006876.hg.1 |
ENST00000533534.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006876.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006876.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006876.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006876.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006876.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006876.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006876.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006876.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006876.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006876.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006876.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006876.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100148422.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100148422.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100148422.hg.1 |
ENST00000256186.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100148422.hg.1 |
NM_032867.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100148422.hg.1 |
TC1100006876.hg.1 |
Clariom D TC |