Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR1600148970.hg.1 |
ENST00000324288.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600148970.hg.1 |
ENST00000381835.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600148970.hg.1 |
ENST00000537380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600148970.hg.1 |
ENST00000570546.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600148970.hg.1 |
ENST00000571186.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600148970.hg.1 |
ENST00000573309.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600148970.hg.1 |
ENST00000576601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600148970.hg.1 |
ENST00000611587.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600148970.hg.1 |
ENST00000618207.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600148970.hg.1 |
ENST00000618327.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600148970.hg.1 |
NM_000246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600148970.hg.1 |
NM_001286402.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600148970.hg.1 |
NM_001286403.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600148970.hg.1 |
NR_104444.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600148970.hg.1 |
ENST00000324288.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600148970.hg.1 |
ENST00000381835.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600148970.hg.1 |
ENST00000537380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600148970.hg.1 |
ENST00000570546.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600148970.hg.1 |
ENST00000571186.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600148970.hg.1 |
ENST00000573309.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600148970.hg.1 |
ENST00000576601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600148970.hg.1 |
ENST00000611587.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600148970.hg.1 |
ENST00000618207.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600148970.hg.1 |
ENST00000618327.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600148970.hg.1 |
NM_000246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600148970.hg.1 |
NM_001286402.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600148970.hg.1 |
NM_001286403.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600148970.hg.1 |
NR_104444.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600148970.hg.1 |
TC1600006888.hg.1 |
Clariom D TC |