Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIITA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000246.3 | NP_000237.2 | 11 - 12 | 2793 | 91 | |
NM_001286402.1 | NP_001273331.1 | 11 - 12 | 2796 | 91 | ||
XM_006720880.3 | XP_006720943.2 | 11 - 12 | 2974 | 91 | ||
XM_011522484.2 | XP_011520786.1 | 11 - 12 | 2974 | 91 | ||
XM_011522485.2 | XP_011520787.1 | 11 - 12 | 2974 | 91 | ||
XM_011522486.2 | XP_011520788.1 | 11 - 12 | 2976 | 91 | ||
XM_011522487.2 | XP_011520789.1 | 10 - 11 | 2711 | 91 | ||
XM_011522488.2 | XP_011520790.1 | 11 - 12 | 4205 | 91 | ||
XM_011522489.2 | XP_011520791.1 | 10 - 11 | 2708 | 91 | ||
XM_011522490.2 | XP_011520792.1 | 11 - 12 | 4203 | 91 | ||
XM_011522491.2 | XP_011520793.1 | 11 - 12 | 2976 | 91 | ||
XM_011522492.2 | XP_011520794.1 | 11 - 12 | 2922 | 91 | ||
XM_011522493.2 | XP_011520795.1 | 11 - 12 | 2919 | 91 | ||
XM_011522494.2 | XP_011520796.1 | 11 - 12 | 2686 | 91 | ||
XM_011522495.2 | XP_011520797.1 | 10 - 11 | 2777 | 91 | ||
XM_011522496.2 | XP_011520798.1 | 10 - 11 | 2774 | 91 | ||
GenBank mRNA | AY699071.1 | - | 10 - 11 | 2591 | 91 | |
U18259.1 | - | 11 - 12 | 2798 | 91 | ||
X74301.1 | - | 11 - 12 | 2775 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Probe spans exons |