Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006756.hg.1 |
ENST00000318881.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006756.hg.1 |
ENST00000318881.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006756.hg.1 |
ENST00000318881.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006756.hg.1 |
ENST00000318881.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006756.hg.1 |
ENST00000318881.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006756.hg.1 |
ENST00000318881.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006756.hg.1 |
ENST00000318881.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006756.hg.1 |
ENST00000318881.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006756.hg.1 |
ENST00000524820.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006756.hg.1 |
ENST00000524820.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006756.hg.1 |
ENST00000524820.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006756.hg.1 |
ENST00000524820.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006756.hg.1 |
ENST00000524820.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006756.hg.1 |
ENST00000524820.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006756.hg.1 |
ENST00000524820.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006756.hg.1 |
ENST00000524820.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006756.hg.1 |
ENST00000524820.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006756.hg.1 |
ENST00000532592.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006756.hg.1 |
ENST00000532592.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006756.hg.1 |
ENST00000532592.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006756.hg.1 |
ENST00000532592.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006756.hg.1 |
ENST00000532592.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006756.hg.1 |
ENST00000532592.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006756.hg.1 |
NM_175733.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006756.hg.1 |
NM_175733.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006756.hg.1 |
NM_175733.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006756.hg.1 |
NM_175733.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006756.hg.1 |
NM_175733.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006756.hg.1 |
NM_175733.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006756.hg.1 |
NM_175733.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006756.hg.1 |
NM_175733.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006756.hg.1 |
ENST00000318881.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006756.hg.1 |
ENST00000318881.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006756.hg.1 |
ENST00000318881.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006756.hg.1 |
ENST00000318881.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006756.hg.1 |
ENST00000318881.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006756.hg.1 |
ENST00000318881.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006756.hg.1 |
ENST00000318881.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006756.hg.1 |
ENST00000318881.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006756.hg.1 |
ENST00000524820.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006756.hg.1 |
ENST00000524820.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006756.hg.1 |
ENST00000524820.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006756.hg.1 |
ENST00000524820.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006756.hg.1 |
ENST00000524820.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006756.hg.1 |
ENST00000524820.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006756.hg.1 |
ENST00000524820.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006756.hg.1 |
ENST00000524820.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006756.hg.1 |
ENST00000524820.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006756.hg.1 |
ENST00000532592.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006756.hg.1 |
ENST00000532592.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006756.hg.1 |
ENST00000532592.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006756.hg.1 |
ENST00000532592.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006756.hg.1 |
ENST00000532592.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006756.hg.1 |
ENST00000532592.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006756.hg.1 |
NM_175733.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006756.hg.1 |
NM_175733.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006756.hg.1 |
NM_175733.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006756.hg.1 |
NM_175733.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006756.hg.1 |
NM_175733.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006756.hg.1 |
NM_175733.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006756.hg.1 |
NM_175733.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006756.hg.1 |
NM_175733.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100147281.hg.1 |
ENST00000318881.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100147281.hg.1 |
ENST00000524820.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100147281.hg.1 |
ENST00000532592.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100147281.hg.1 |
NM_175733.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100147281.hg.1 |
ENST00000318881.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100147281.hg.1 |
ENST00000524820.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100147281.hg.1 |
ENST00000532592.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100147281.hg.1 |
NM_175733.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100147281.hg.1 |
TC1100006756.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100147277.hg.1 |
ENST00000524820.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100147277.hg.1 |
ENST00000524820.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100147277.hg.1 |
TC1100006756.hg.1 |
Clariom D TC |