Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024901.4 | NP_079177.2 | 1 - 2 | 310 | 68 | |
XM_006710922.1 | XP_006710985.1 | 1 - 2 | 254 | 68 | ||
GenBank mRNA | AL713773.1 | - | 1 - 2 | 153 | 68 | |
BC004557.2 | - | 1 - 2 | 266 | 3957641 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |