Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR1500182193.hg.1 |
ENST00000329464.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182193.hg.1 |
ENST00000338264.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182193.hg.1 |
ENST00000558173.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182193.hg.1 |
ENST00000558329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182193.hg.1 |
ENST00000560442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182193.hg.1 |
ENST00000561170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182193.hg.1 |
NM_001301144.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500182193.hg.1 |
NM_001301145.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500182193.hg.1 |
NM_080745.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500182193.hg.1 |
NM_182985.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500182193.hg.1 |
ENST00000329464.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182193.hg.1 |
ENST00000338264.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182193.hg.1 |
ENST00000558173.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182193.hg.1 |
ENST00000558329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182193.hg.1 |
ENST00000560442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182193.hg.1 |
ENST00000561170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182193.hg.1 |
NM_001301144.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500182193.hg.1 |
NM_001301145.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500182193.hg.1 |
NM_080745.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500182193.hg.1 |
NM_182985.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500182193.hg.1 |
TC1500010732.hg.1 |
Clariom D TC |