Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009689.hg.1 |
ENST00000442234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009689.hg.1 |
NM_206937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009689.hg.1 |
TC13000867.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009686.hg.1 |
ENST00000356922.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009686.hg.1 |
NM_002312.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009686.hg.1 |
NM_206937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009686.hg.1 |
TC13000867.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
ENST00000356922.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
ENST00000356922.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
ENST00000356922.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
ENST00000356922.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
ENST00000356922.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
ENST00000356922.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
ENST00000405925.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
ENST00000405925.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
ENST00000405925.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
ENST00000405925.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
ENST00000405925.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
ENST00000442234.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
ENST00000442234.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
ENST00000442234.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
ENST00000442234.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
ENST00000442234.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
ENST00000442234.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
ENST00000611712.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
ENST00000611712.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
ENST00000611712.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
ENST00000611712.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
ENST00000611712.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
ENST00000611712.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
ENST00000611712.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
ENST00000611712.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
ENST00000614526.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
ENST00000614526.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
ENST00000614526.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
ENST00000614526.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
NM_001098268.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
NM_001098268.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
NM_001098268.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
NM_001098268.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
NM_001098268.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
NM_002312.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
NM_002312.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
NM_002312.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
NM_002312.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
NM_002312.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
NM_002312.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
NM_206937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
NM_206937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
NM_206937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
NM_206937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
NM_206937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009740.hg.1 |
NM_206937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300164995.hg.1 |
ENST00000356922.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300164995.hg.1 |
ENST00000442234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300164995.hg.1 |
ENST00000611712.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300164995.hg.1 |
ENST00000614526.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300164995.hg.1 |
NM_002312.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300164995.hg.1 |
NM_206937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300164995.hg.1 |
ENST00000356922.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300164995.hg.1 |
ENST00000442234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300164995.hg.1 |
ENST00000611712.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300164995.hg.1 |
ENST00000614526.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300164995.hg.1 |
NM_002312.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300164995.hg.1 |
NM_206937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300164995.hg.1 |
TC1300009740.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300164998.hg.1 |
ENST00000442234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300164998.hg.1 |
ENST00000611712.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300164998.hg.1 |
ENST00000614526.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300164998.hg.1 |
NM_206937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300164998.hg.1 |
ENST00000442234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300164998.hg.1 |
ENST00000611712.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300164998.hg.1 |
ENST00000614526.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300164998.hg.1 |
NM_206937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300164998.hg.1 |
TC1300009740.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
ENST00000356922.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
ENST00000356922.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
ENST00000356922.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
ENST00000356922.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
ENST00000356922.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
ENST00000356922.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
ENST00000405925.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
ENST00000405925.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
ENST00000405925.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
ENST00000405925.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
ENST00000405925.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
ENST00000442234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
ENST00000442234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
ENST00000442234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
ENST00000442234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
ENST00000442234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
ENST00000442234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
ENST00000611712.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
ENST00000611712.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
ENST00000611712.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
ENST00000611712.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
ENST00000611712.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
ENST00000611712.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
ENST00000611712.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
ENST00000611712.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
ENST00000614526.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
ENST00000614526.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
ENST00000614526.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
ENST00000614526.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
NM_001098268.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
NM_001098268.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
NM_001098268.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
NM_001098268.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
NM_001098268.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
NM_002312.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
NM_002312.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
NM_002312.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
NM_002312.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
NM_002312.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
NM_002312.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
NM_206937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
NM_206937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
NM_206937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
NM_206937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
NM_206937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009740.hg.1 |
NM_206937.1 |
Refseq |