Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
ENST00000311461.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
ENST00000311461.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
ENST00000311461.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
ENST00000311461.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
ENST00000311461.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
ENST00000311461.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
ENST00000311461.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
ENST00000311461.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
ENST00000311461.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
ENST00000311461.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
ENST00000311469.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
ENST00000311469.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
ENST00000311469.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
ENST00000311469.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
ENST00000311469.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
ENST00000311469.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
ENST00000311469.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
ENST00000311469.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
ENST00000311469.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
ENST00000311469.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
ENST00000311469.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
ENST00000503391.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
ENST00000503391.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
ENST00000503391.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
ENST00000503391.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
ENST00000503391.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
ENST00000503391.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
ENST00000503391.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
ENST00000503391.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
ENST00000503391.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
ENST00000503915.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
ENST00000503915.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
ENST00000503915.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
ENST00000503915.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
ENST00000503915.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
ENST00000503915.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
ENST00000503915.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
ENST00000503915.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
ENST00000503915.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
ENST00000514935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
ENST00000514935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
ENST00000514935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
ENST00000514935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
NM_015697.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
NM_015697.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
NM_015697.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
NM_015697.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
NM_015697.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
NM_015697.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
NM_015697.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
NM_015697.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
NM_015697.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
NM_015697.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011202.hg.1 |
NM_015697.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04018663.hg.1 |
TC04001346.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04018663.hg.1 |
ENST00000311461.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04018663.hg.1 |
ENST00000311469.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04018663.hg.1 |
ENST00000439031.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04018663.hg.1 |
NM_015697.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
ENST00000311461.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
ENST00000311461.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
ENST00000311461.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
ENST00000311461.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
ENST00000311461.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
ENST00000311461.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
ENST00000311461.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
ENST00000311461.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
ENST00000311461.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
ENST00000311461.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
ENST00000311469.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
ENST00000311469.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
ENST00000311469.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
ENST00000311469.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
ENST00000311469.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
ENST00000311469.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
ENST00000311469.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
ENST00000311469.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
ENST00000311469.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
ENST00000311469.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
ENST00000311469.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
ENST00000503391.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
ENST00000503391.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
ENST00000503391.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
ENST00000503391.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
ENST00000503391.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
ENST00000503391.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
ENST00000503391.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
ENST00000503391.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
ENST00000503391.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
ENST00000503915.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
ENST00000503915.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
ENST00000503915.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
ENST00000503915.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
ENST00000503915.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
ENST00000503915.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
ENST00000503915.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
ENST00000503915.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
ENST00000503915.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
ENST00000514935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
ENST00000514935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
ENST00000514935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
ENST00000514935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
NM_015697.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
NM_015697.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
NM_015697.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
NM_015697.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
NM_015697.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
NM_015697.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
NM_015697.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
NM_015697.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
NM_015697.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
NM_015697.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011202.hg.1 |
NM_015697.7 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400176431.hg.1 |
ENST00000311461.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400176431.hg.1 |
ENST00000311469.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400176431.hg.1 |
ENST00000503915.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400176431.hg.1 |
NM_015697.7 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400176431.hg.1 |
ENST00000311461.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400176431.hg.1 |
ENST00000311469.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400176431.hg.1 |
ENST00000503915.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400176431.hg.1 |
NM_015697.7 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400176431.hg.1 |
TC0400011202.hg.1 |
Clariom D TC |