Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC42BPG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017525.2 | NP_059995.2 | 29 - 30 | 3365 | 147 | |
XM_011545155.2 | XP_011543457.1 | 29 - 30 | 3534 | 147 | ||
XM_011545156.2 | XP_011543458.1 | 29 - 30 | 3514 | 147 | ||
XM_011545157.2 | XP_011543459.1 | 29 - 30 | 3535 | 147 | ||
XM_011545158.2 | XP_011543460.1 | 29 - 30 | 3535 | 147 | ||
XM_011545159.2 | XP_011543461.1 | 29 - 30 | 3535 | 147 | ||
XM_011545160.2 | XP_011543462.1 | 30 - 31 | 3486 | 147 | ||
XM_017017996.1 | XP_016873485.1 | 29 - 30 | 3531 | 147 | ||
XM_017017998.1 | XP_016873487.1 | 28 - 29 | 3318 | 147 | ||
XM_017017999.1 | XP_016873488.1 | 19 - 20 | 2114 | 147 | ||
GenBank mRNA | AY648038.1 | - | 29 - 30 | 3365 | 147 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase 3' Most Detect Genomic DNA Homo sapiens Probe spans exons |