Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031309.5 | NP_112599.2 | 2 | 3341 | 105 | |
GenBank mRNA | AL512683.1 | - | 1 | 856 | 105 | |
BC024967.2 | - | 1 | 862 | 105 | ||
BC093806.1 | - | 1 | 299 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |