Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023792.hg.1 |
TC06001941.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023792.hg.1 |
ENST00000359203.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023792.hg.1 |
ENST00000369415.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023792.hg.1 |
NM_021244.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
ENST00000359203.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
ENST00000359203.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
ENST00000359203.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
ENST00000359203.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
ENST00000359203.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
ENST00000359203.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
ENST00000359203.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
ENST00000359203.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
ENST00000359203.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
ENST00000359203.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
ENST00000359203.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
ENST00000369415.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
ENST00000369415.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
ENST00000369415.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
ENST00000369415.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
ENST00000369415.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
ENST00000369415.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
ENST00000369415.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
ENST00000369415.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
ENST00000369415.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
ENST00000369415.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
ENST00000369415.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
ENST00000369415.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
ENST00000369415.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
ENST00000369415.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
ENST00000492783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
ENST00000492783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
ENST00000492783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
ENST00000492783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
ENST00000492783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
NM_021244.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
NM_021244.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
NM_021244.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
NM_021244.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
NM_021244.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
NM_021244.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
NM_021244.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
NM_021244.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
NM_021244.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
NM_021244.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
NM_021244.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
NM_021244.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
NM_021244.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012544.hg.1 |
NM_021244.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023793.hg.1 |
TC06001941.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023793.hg.1 |
ENST00000359203.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023793.hg.1 |
ENST00000369415.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023793.hg.1 |
NM_021244.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186442.hg.1 |
ENST00000359203.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186442.hg.1 |
ENST00000369415.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186442.hg.1 |
NM_021244.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186442.hg.1 |
ENST00000359203.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186442.hg.1 |
ENST00000369415.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186442.hg.1 |
NM_021244.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186442.hg.1 |
TC0600012544.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600186441.hg.1 |
ENST00000359203.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186441.hg.1 |
ENST00000369415.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186441.hg.1 |
NM_021244.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186441.hg.1 |
ENST00000359203.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186441.hg.1 |
ENST00000369415.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186441.hg.1 |
NM_021244.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186441.hg.1 |
TC0600012544.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
ENST00000359203.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
ENST00000359203.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
ENST00000359203.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
ENST00000359203.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
ENST00000359203.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
ENST00000359203.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
ENST00000359203.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
ENST00000359203.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
ENST00000359203.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
ENST00000359203.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
ENST00000359203.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
ENST00000369415.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
ENST00000369415.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
ENST00000369415.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
ENST00000369415.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
ENST00000369415.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
ENST00000369415.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
ENST00000369415.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
ENST00000369415.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
ENST00000369415.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
ENST00000369415.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
ENST00000369415.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
ENST00000369415.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
ENST00000369415.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
ENST00000369415.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
ENST00000492783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
ENST00000492783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
ENST00000492783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
ENST00000492783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
ENST00000492783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
NM_021244.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
NM_021244.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
NM_021244.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
NM_021244.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
NM_021244.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
NM_021244.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
NM_021244.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
NM_021244.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
NM_021244.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
NM_021244.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
NM_021244.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
NM_021244.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
NM_021244.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012544.hg.1 |
NM_021244.4 |
Refseq |