Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000322247.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000322247.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000322247.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000322247.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000322247.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000322247.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000322247.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000322247.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000322247.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000322247.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000322247.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000322247.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000322247.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000402563.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000402563.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000402563.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000402563.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000402563.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000402563.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000402563.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000402563.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000402563.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000402563.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000402563.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000402563.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000402563.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000585785.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000585785.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000585785.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000585785.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000585785.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000585785.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000585785.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000588475.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000588475.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000588475.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000588475.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000588475.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000588475.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000589346.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000589346.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000589346.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000589346.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000589346.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000591269.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000591269.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000591269.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000591269.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000591269.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000591269.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000591269.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000591269.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000591269.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000592976.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000592976.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000592976.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000592976.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000592976.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000592976.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000592976.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000592976.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
ENST00000592976.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
NM_001098801.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
NM_001098801.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
NM_001098801.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
NM_001098801.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
NM_001098801.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
NM_001098801.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
NM_001098801.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
NM_001098801.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
NM_001098801.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
NM_001098801.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
NM_001098801.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
NM_001098801.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
NM_001098801.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
NM_001098801.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
NM_152352.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
NM_152352.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
NM_152352.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
NM_152352.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
NM_152352.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
NM_152352.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
NM_152352.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
NM_152352.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
NM_152352.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
NM_152352.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
NM_152352.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
NM_152352.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008162.hg.1 |
NM_152352.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800155798.hg.1 |
ENST00000322247.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800155798.hg.1 |
ENST00000402563.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800155798.hg.1 |
ENST00000589346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800155798.hg.1 |
ENST00000591269.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800155798.hg.1 |
ENST00000592976.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800155798.hg.1 |
NM_001098801.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800155798.hg.1 |
NM_152352.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800155798.hg.1 |
TC1800008162.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800155798.hg.1 |
ENST00000322247.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800155798.hg.1 |
ENST00000402563.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800155798.hg.1 |
ENST00000589346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800155798.hg.1 |
ENST00000591269.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800155798.hg.1 |
ENST00000592976.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800155798.hg.1 |
NM_001098801.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800155798.hg.1 |
NM_152352.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800155797.hg.1 |
ENST00000322247.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800155797.hg.1 |
ENST00000402563.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800155797.hg.1 |
ENST00000589346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800155797.hg.1 |
ENST00000592976.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800155797.hg.1 |
NM_001098801.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800155797.hg.1 |
NM_152352.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800155797.hg.1 |
TC1800008162.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800155797.hg.1 |
ENST00000322247.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800155797.hg.1 |
ENST00000402563.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800155797.hg.1 |
ENST00000589346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800155797.hg.1 |
ENST00000592976.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800155797.hg.1 |
NM_001098801.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800155797.hg.1 |
NM_152352.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000322247.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000322247.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000322247.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000322247.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000322247.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000322247.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000322247.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000322247.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000322247.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000322247.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000322247.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000322247.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000322247.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000402563.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000402563.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000402563.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000402563.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000402563.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000402563.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000402563.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000402563.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000402563.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000402563.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000402563.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000402563.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000402563.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000585785.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000585785.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000585785.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000585785.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000585785.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000585785.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000585785.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000588475.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000588475.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000588475.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000588475.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000588475.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000588475.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000589346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000589346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000589346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000589346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000589346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000591269.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000591269.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000591269.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000591269.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000591269.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000591269.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000591269.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000591269.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000591269.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000592976.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000592976.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000592976.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000592976.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000592976.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000592976.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000592976.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000592976.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
ENST00000592976.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
NM_001098801.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
NM_001098801.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
NM_001098801.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
NM_001098801.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
NM_001098801.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
NM_001098801.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
NM_001098801.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
NM_001098801.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
NM_001098801.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
NM_001098801.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
NM_001098801.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
NM_001098801.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
NM_001098801.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
NM_001098801.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
NM_152352.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
NM_152352.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
NM_152352.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
NM_152352.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
NM_152352.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
NM_152352.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
NM_152352.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
NM_152352.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
NM_152352.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
NM_152352.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
NM_152352.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
NM_152352.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008162.hg.1 |
NM_152352.3 |
Refseq |