Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024293.hg.1 |
TC04001761.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024293.hg.1 |
ENST00000280191.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024293.hg.1 |
ENST00000515234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024293.hg.1 |
NM_144644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012477.hg.1 |
ENST00000280191.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012477.hg.1 |
ENST00000280191.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012477.hg.1 |
ENST00000280191.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012477.hg.1 |
ENST00000280191.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012477.hg.1 |
ENST00000280191.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012477.hg.1 |
ENST00000280191.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012477.hg.1 |
ENST00000280191.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012477.hg.1 |
ENST00000280191.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012477.hg.1 |
ENST00000280191.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012477.hg.1 |
ENST00000508699.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012477.hg.1 |
ENST00000508699.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012477.hg.1 |
ENST00000508699.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012477.hg.1 |
ENST00000515234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012477.hg.1 |
ENST00000515234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012477.hg.1 |
ENST00000515234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012477.hg.1 |
ENST00000515234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012477.hg.1 |
ENST00000515234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012477.hg.1 |
ENST00000515234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012477.hg.1 |
ENST00000515234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012477.hg.1 |
ENST00000515234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012477.hg.1 |
NM_144644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012477.hg.1 |
NM_144644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012477.hg.1 |
NM_144644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012477.hg.1 |
NM_144644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012477.hg.1 |
NM_144644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012477.hg.1 |
NM_144644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012477.hg.1 |
NM_144644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012477.hg.1 |
NM_144644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012477.hg.1 |
NM_144644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012477.hg.1 |
NM_144644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185558.hg.1 |
ENST00000280191.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185558.hg.1 |
ENST00000508699.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185558.hg.1 |
ENST00000515234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185558.hg.1 |
NM_144644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185558.hg.1 |
ENST00000280191.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185558.hg.1 |
ENST00000508699.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185558.hg.1 |
ENST00000515234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185558.hg.1 |
NM_144644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185558.hg.1 |
TC0400012477.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400012477.hg.1 |
ENST00000280191.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012477.hg.1 |
ENST00000280191.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012477.hg.1 |
ENST00000280191.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012477.hg.1 |
ENST00000280191.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012477.hg.1 |
ENST00000280191.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012477.hg.1 |
ENST00000280191.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012477.hg.1 |
ENST00000280191.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012477.hg.1 |
ENST00000280191.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012477.hg.1 |
ENST00000280191.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012477.hg.1 |
ENST00000508699.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012477.hg.1 |
ENST00000508699.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012477.hg.1 |
ENST00000508699.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012477.hg.1 |
ENST00000515234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012477.hg.1 |
ENST00000515234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012477.hg.1 |
ENST00000515234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012477.hg.1 |
ENST00000515234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012477.hg.1 |
ENST00000515234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012477.hg.1 |
ENST00000515234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012477.hg.1 |
ENST00000515234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012477.hg.1 |
ENST00000515234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012477.hg.1 |
NM_144644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012477.hg.1 |
NM_144644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012477.hg.1 |
NM_144644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012477.hg.1 |
NM_144644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012477.hg.1 |
NM_144644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012477.hg.1 |
NM_144644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012477.hg.1 |
NM_144644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012477.hg.1 |
NM_144644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012477.hg.1 |
NM_144644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012477.hg.1 |
NM_144644.3 |
Refseq |