Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012293.hg.1 |
ENST00000370315.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012293.hg.1 |
ENST00000370315.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012293.hg.1 |
ENST00000370315.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012293.hg.1 |
ENST00000370315.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012293.hg.1 |
ENST00000370315.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012293.hg.1 |
ENST00000370315.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012293.hg.1 |
ENST00000370315.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012293.hg.1 |
ENST00000370315.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012293.hg.1 |
ENST00000370315.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012293.hg.1 |
ENST00000370315.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012293.hg.1 |
ENST00000370315.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012293.hg.1 |
ENST00000370315.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012293.hg.1 |
ENST00000370318.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012293.hg.1 |
ENST00000370318.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012293.hg.1 |
ENST00000370318.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012293.hg.1 |
ENST00000370318.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012293.hg.1 |
ENST00000370318.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012293.hg.1 |
ENST00000370318.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012293.hg.1 |
ENST00000370318.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012293.hg.1 |
ENST00000370318.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012293.hg.1 |
ENST00000459924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012293.hg.1 |
ENST00000459924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012293.hg.1 |
ENST00000459924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012293.hg.1 |
NM_138441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012293.hg.1 |
NM_138441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012293.hg.1 |
NM_138441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012293.hg.1 |
NM_138441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012293.hg.1 |
NM_138441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012293.hg.1 |
NM_138441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012293.hg.1 |
NM_138441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012293.hg.1 |
NM_138441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012293.hg.1 |
NM_138441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012293.hg.1 |
NM_138441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012293.hg.1 |
NM_138441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022695.hg.1 |
TC06001873.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022695.hg.1 |
ENST00000296913.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022695.hg.1 |
ENST00000370315.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022695.hg.1 |
ENST00000370318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022695.hg.1 |
NM_138441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012293.hg.1 |
ENST00000370315.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012293.hg.1 |
ENST00000370315.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012293.hg.1 |
ENST00000370315.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012293.hg.1 |
ENST00000370315.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012293.hg.1 |
ENST00000370315.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012293.hg.1 |
ENST00000370315.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012293.hg.1 |
ENST00000370315.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012293.hg.1 |
ENST00000370315.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012293.hg.1 |
ENST00000370315.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012293.hg.1 |
ENST00000370315.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012293.hg.1 |
ENST00000370315.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012293.hg.1 |
ENST00000370315.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012293.hg.1 |
ENST00000370318.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012293.hg.1 |
ENST00000370318.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012293.hg.1 |
ENST00000370318.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012293.hg.1 |
ENST00000370318.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012293.hg.1 |
ENST00000370318.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012293.hg.1 |
ENST00000370318.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012293.hg.1 |
ENST00000370318.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012293.hg.1 |
ENST00000370318.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012293.hg.1 |
ENST00000459924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012293.hg.1 |
ENST00000459924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012293.hg.1 |
ENST00000459924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012293.hg.1 |
NM_138441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012293.hg.1 |
NM_138441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012293.hg.1 |
NM_138441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012293.hg.1 |
NM_138441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012293.hg.1 |
NM_138441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012293.hg.1 |
NM_138441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012293.hg.1 |
NM_138441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012293.hg.1 |
NM_138441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012293.hg.1 |
NM_138441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012293.hg.1 |
NM_138441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012293.hg.1 |
NM_138441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600184711.hg.1 |
ENST00000370315.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600184711.hg.1 |
ENST00000370318.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600184711.hg.1 |
NM_138441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600184711.hg.1 |
ENST00000370315.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600184711.hg.1 |
ENST00000370318.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600184711.hg.1 |
NM_138441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600184711.hg.1 |
TC0600012293.hg.1 |
Clariom D TC |