Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019469.hg.1 |
ENST00000254051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019469.hg.1 |
ENST00000377816.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019469.hg.1 |
ENST00000394072.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019469.hg.1 |
NM_032865.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019469.hg.1 |
TC17001465.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
ENST00000254051.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
ENST00000254051.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
ENST00000254051.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
ENST00000254051.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
ENST00000254051.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
ENST00000254051.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
ENST00000254051.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
ENST00000254051.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
ENST00000254051.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
ENST00000254051.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
ENST00000254051.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
ENST00000254051.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
ENST00000254051.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
ENST00000254051.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
ENST00000254051.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
ENST00000254051.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
ENST00000394072.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
ENST00000394072.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
ENST00000394072.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
ENST00000394072.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
ENST00000394072.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
ENST00000394072.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
ENST00000497303.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
ENST00000497303.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
ENST00000497303.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
ENST00000497303.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
ENST00000497303.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
ENST00000582747.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
ENST00000582747.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
ENST00000582747.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
ENST00000582747.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
NM_032865.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
NM_032865.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
NM_032865.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
NM_032865.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
NM_032865.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
NM_032865.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
NM_032865.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
NM_032865.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
NM_032865.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
NM_032865.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
NM_032865.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
NM_032865.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
NM_032865.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
NM_032865.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
NM_032865.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010625.hg.1 |
NM_032865.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700183828.hg.1 |
ENST00000254051.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700183828.hg.1 |
ENST00000394072.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700183828.hg.1 |
NM_032865.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700183828.hg.1 |
ENST00000254051.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700183828.hg.1 |
ENST00000394072.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700183828.hg.1 |
NM_032865.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700183828.hg.1 |
TC1700010625.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
ENST00000254051.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
ENST00000254051.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
ENST00000254051.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
ENST00000254051.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
ENST00000254051.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
ENST00000254051.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
ENST00000254051.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
ENST00000254051.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
ENST00000254051.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
ENST00000254051.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
ENST00000254051.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
ENST00000254051.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
ENST00000254051.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
ENST00000254051.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
ENST00000254051.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
ENST00000254051.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
ENST00000394072.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
ENST00000394072.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
ENST00000394072.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
ENST00000394072.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
ENST00000394072.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
ENST00000394072.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
ENST00000497303.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
ENST00000497303.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
ENST00000497303.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
ENST00000497303.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
ENST00000497303.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
ENST00000582747.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
ENST00000582747.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
ENST00000582747.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
ENST00000582747.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
NM_032865.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
NM_032865.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
NM_032865.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
NM_032865.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
NM_032865.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
NM_032865.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
NM_032865.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
NM_032865.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
NM_032865.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
NM_032865.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
NM_032865.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
NM_032865.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
NM_032865.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
NM_032865.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
NM_032865.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010625.hg.1 |
NM_032865.5 |
Refseq |