Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008489.hg.1 |
ENST00000222157.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008489.hg.1 |
ENST00000222157.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008489.hg.1 |
ENST00000222157.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008489.hg.1 |
ENST00000222157.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008489.hg.1 |
ENST00000222157.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008489.hg.1 |
ENST00000593756.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008489.hg.1 |
ENST00000593756.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008489.hg.1 |
ENST00000593756.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008489.hg.1 |
ENST00000593756.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008489.hg.1 |
ENST00000593756.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008489.hg.1 |
ENST00000593756.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008489.hg.1 |
ENST00000593756.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008489.hg.1 |
NM_019113.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008489.hg.1 |
NM_019113.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008489.hg.1 |
NM_019113.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008489.hg.1 |
NM_019113.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008489.hg.1 |
NM_019113.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900165369.hg.1 |
ENST00000222157.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165369.hg.1 |
ENST00000593756.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165369.hg.1 |
NM_019113.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900165369.hg.1 |
TC1900008489.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900165369.hg.1 |
ENST00000222157.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165369.hg.1 |
ENST00000593756.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165369.hg.1 |
NM_019113.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008489.hg.1 |
ENST00000222157.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008489.hg.1 |
ENST00000222157.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008489.hg.1 |
ENST00000222157.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008489.hg.1 |
ENST00000222157.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008489.hg.1 |
ENST00000222157.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008489.hg.1 |
ENST00000593756.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008489.hg.1 |
ENST00000593756.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008489.hg.1 |
ENST00000593756.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008489.hg.1 |
ENST00000593756.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008489.hg.1 |
ENST00000593756.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008489.hg.1 |
ENST00000593756.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008489.hg.1 |
ENST00000593756.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008489.hg.1 |
NM_019113.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008489.hg.1 |
NM_019113.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008489.hg.1 |
NM_019113.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008489.hg.1 |
NM_019113.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008489.hg.1 |
NM_019113.3 |
Refseq |