Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08021199.hg.1 |
TC08001637.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08021199.hg.1 |
ENST00000276708.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08021199.hg.1 |
NM_031415.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08021198.hg.1 |
TC08001637.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08021198.hg.1 |
ENST00000276708.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08021198.hg.1 |
NM_031415.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000276708.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000276708.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000276708.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000276708.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000276708.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000276708.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000276708.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000276708.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000276708.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000276708.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000276708.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000276708.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000276708.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000276708.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000276708.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000276708.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000276708.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000521365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000521365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000521365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000521365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000522273.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000522273.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000522273.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000522273.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000522273.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000522273.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000522273.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000522273.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000522273.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000522273.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000522273.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000619643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000619643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000619643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000619643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000619643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000619643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000619643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000619643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000619643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000619643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000619643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000619643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000619643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000619643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
ENST00000619643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
NM_031415.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
NM_031415.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
NM_031415.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
NM_031415.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
NM_031415.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
NM_031415.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
NM_031415.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
NM_031415.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
NM_031415.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
NM_031415.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
NM_031415.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
NM_031415.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
NM_031415.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
NM_031415.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
NM_031415.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
NM_031415.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011814.hg.1 |
NM_031415.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08021196.hg.1 |
TC08001637.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08021196.hg.1 |
ENST00000276708.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08021196.hg.1 |
NM_031415.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800180174.hg.1 |
ENST00000276708.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800180174.hg.1 |
ENST00000521365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800180174.hg.1 |
ENST00000522273.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800180174.hg.1 |
ENST00000619643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800180174.hg.1 |
NM_031415.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800180174.hg.1 |
ENST00000276708.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800180174.hg.1 |
ENST00000521365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800180174.hg.1 |
ENST00000522273.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800180174.hg.1 |
ENST00000619643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800180174.hg.1 |
NM_031415.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800180174.hg.1 |
TC0800011814.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000276708.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000276708.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000276708.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000276708.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000276708.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000276708.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000276708.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000276708.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000276708.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000276708.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000276708.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000276708.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000276708.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000276708.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000276708.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000276708.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000276708.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000521365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000521365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000521365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000521365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000522273.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000522273.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000522273.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000522273.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000522273.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000522273.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000522273.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000522273.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000522273.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000522273.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000522273.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000619643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000619643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000619643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000619643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000619643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000619643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000619643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000619643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000619643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000619643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000619643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000619643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000619643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000619643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
ENST00000619643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
NM_031415.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
NM_031415.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
NM_031415.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
NM_031415.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
NM_031415.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
NM_031415.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
NM_031415.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
NM_031415.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
NM_031415.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
NM_031415.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
NM_031415.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
NM_031415.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
NM_031415.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
NM_031415.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
NM_031415.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
NM_031415.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011814.hg.1 |
NM_031415.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800180171.hg.1 |
ENST00000276708.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800180171.hg.1 |
ENST00000522273.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800180171.hg.1 |
ENST00000619643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800180171.hg.1 |
NM_031415.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800180171.hg.1 |
ENST00000276708.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800180171.hg.1 |
ENST00000522273.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800180171.hg.1 |
ENST00000619643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800180171.hg.1 |
NM_031415.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800180171.hg.1 |
TC0800011814.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800180173.hg.1 |
ENST00000276708.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800180173.hg.1 |
ENST00000521365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800180173.hg.1 |
ENST00000522273.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800180173.hg.1 |
ENST00000619643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800180173.hg.1 |
NM_031415.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800180173.hg.1 |
ENST00000276708.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800180173.hg.1 |
ENST00000521365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800180173.hg.1 |
ENST00000522273.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800180173.hg.1 |
ENST00000619643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800180173.hg.1 |
NM_031415.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800180173.hg.1 |
TC0800011814.hg.1 |
Clariom D TC |