Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04635288 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24825779 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24825724 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24825766 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24825750 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04635304 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04635251 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24825743 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24825787 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24825777 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24825764 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24825763 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24825715 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24825755 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04635292 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24825735 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04635266 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24825761 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24825781 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24825765 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24825727 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24825713 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24825752 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04635295 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24825707 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24825772 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04635296 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24825739 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04635273 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24825733 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24825725 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04635249 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04635271 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24825723 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04635264 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24825706 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24825737 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04635263 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24825717 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24825738 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24825770 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04635269 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24825782 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24825767 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24825759 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24825756 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24825716 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24825769 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24825726 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04635281 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24825742 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15797420 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24825746 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24825785 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04635294 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24825757 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04635231 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24825718 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24825708 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
180149 |
NM_020795.2 |
refseq_mrna_human_NM |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
262337 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
127340 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
3236 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
233026 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3313366 |
- |
- |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_HS_44k_0586 |
- |
- |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_HS_44k_0587 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
23065 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P31709774 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3313366 |
- |
- |
SurePrint G3 Human GE Microarray G4851A |
A_33_P3313366 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825757 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825779 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825743 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825769 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825752 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04635292 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04635264 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825777 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825789 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04635285 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825746 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04635294 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825772 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825742 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04635271 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825755 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04635296 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04635266 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04635295 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825793 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825750 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825764 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04635300 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825766 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825785 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825756 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825759 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825761 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04635288 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825763 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825781 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825770 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825794 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825787 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04635281 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04635304 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825783 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04635273 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825782 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825779 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04635264 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825750 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825772 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825782 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825755 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825787 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04635273 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04635285 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04635294 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825746 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825759 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825752 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825763 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825743 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825783 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825756 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825793 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825769 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04635266 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825761 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825764 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04635271 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04635300 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04635292 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825770 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04635288 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04635304 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825777 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04635281 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825766 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825794 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825781 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04635295 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825785 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04635296 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825789 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825742 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24825757 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P04635292 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P04635259 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P04635305 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P04635262 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P04635299 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P04635290 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P04635287 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P04635302 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P04635281 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P04635261 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P04635303 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P04635274 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P04635301 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P04635266 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P04635279 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P04635271 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P04635297 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P04635280 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P04635277 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P04635285 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P04635293 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P04635268 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P04635264 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P04635283 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P04635295 |
- |
- |
Agilent Human 1B Microarray G4111A |
A_32_P361660 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15797420 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04635299 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04635295 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04635277 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04635247 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04635245 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04635270 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04635281 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04635261 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04635274 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04635292 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04635271 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04635238 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04635257 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04635231 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04635241 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04635290 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04635250 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04635239 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15797424 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04635268 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04635255 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04635236 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04635285 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04635283 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04635244 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04635287 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04635259 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04635279 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04635264 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15797422 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P17492451 |
- |
- |