Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08001141.hg.1 |
TC08000125.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08001141.hg.1 |
ENST00000262096.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08001141.hg.1 |
NM_016353.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
ENST00000517334.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
ENST00000517334.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
ENST00000517334.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
ENST00000522184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
ENST00000522184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
ENST00000522184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
ENST00000522184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
ENST00000522184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
ENST00000522184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
ENST00000522184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
ENST00000523132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
ENST00000523132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
NM_016353.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
NM_016353.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
NM_016353.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
NM_016353.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
NM_016353.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
NM_016353.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
NM_016353.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
NM_016353.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
NM_016353.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
NM_016353.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
NM_016353.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
NM_016353.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
NM_016353.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
NM_016353.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
NM_016353.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
NM_016353.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
NM_016353.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
NM_016353.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006864.hg.1 |
NM_016353.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800146425.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800146425.hg.1 |
NM_016353.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800146425.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800146425.hg.1 |
NM_016353.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800146425.hg.1 |
TC0800006864.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
ENST00000262096.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
ENST00000517334.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
ENST00000517334.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
ENST00000517334.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
ENST00000522184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
ENST00000522184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
ENST00000522184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
ENST00000522184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
ENST00000522184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
ENST00000522184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
ENST00000522184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
ENST00000523132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
ENST00000523132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
NM_016353.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
NM_016353.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
NM_016353.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
NM_016353.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
NM_016353.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
NM_016353.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
NM_016353.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
NM_016353.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
NM_016353.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
NM_016353.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
NM_016353.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
NM_016353.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
NM_016353.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
NM_016353.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
NM_016353.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
NM_016353.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
NM_016353.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
NM_016353.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006864.hg.1 |
NM_016353.4 |
Refseq |