Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs00938197_m1
See other DLG1 GE Assays ›
Gene Symbol:
DLG1
Gene Name:
discs large MAGUK scaffold protein 1
Gene Aliases:
DLGH1, SAP-97, SAP97, dJ1061C18.1.1, hdlg
Chromosome Location:
Chr.3: 197042560 - 197299272 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs00938197_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: DLG1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001204386.1
NP_001191315.1
18 - 19
2192
81
NM_004087.2
NP_004078.2
19 - 20
2294
81
XM_005269289.3
XP_005269346.1
19 - 20
2576
81
XM_011512503.1
XP_011510805.1
18 - 19
2471
81
XM_017005800.1
XP_016861289.1
19 - 20
2711
81
XM_017005801.1
XP_016861290.1
19 - 20
2231
81
XM_017005802.1
XP_016861291.1
19 - 20
2301
81
XM_017005803.1
XP_016861292.1
19 - 20
2222
81
XM_017005804.1
XP_016861293.1
19 - 20
2573
81
XM_017005806.1
XP_016861295.1
18 - 19
2474
81
XM_017005807.1
XP_016861296.1
18 - 19
2573
81
XM_017005808.1
XP_016861297.1
18 - 19
2132
81
XM_017005809.1
XP_016861298.1
18 - 19
2234
81
XM_017005810.1
XP_016861299.1
18 - 19
2129
81
XM_017005811.1
XP_016861300.1
17 - 18
2420
81
XM_017005812.1
XP_016861301.1
17 - 18
2074
81
XM_017005813.1
XP_016861302.1
17 - 18
2078
81
XM_017005814.1
XP_016861303.1
17 - 18
2181
81
XM_017005815.1
XP_016861304.1
17 - 18
2417
81
XM_017005821.1
XP_016861310.1
15 - 16
4579
81
XM_017005823.1
XP_016861312.1
14 - 15
4476
81
XM_017005824.1
XP_016861313.1
9 - 10
1095
81
GenBank mRNA
BC140841.1
-
19 - 20
2259
9021709
81
BC144651.1
-
18 - 19
2157
9053182
81
U13896.1
-
19 - 20
2292
81
Back To Top
More Information
Set Membership:
Ambion siRNA available
Amplicon length between 71 and 85
Mammalian Gene Collection
Homo sapiens
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene DLG1
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
branching involved in ureteric bud morphogenesis
immunological synapse formation
endothelial cell proliferation
lens development in camera-type eye
T cell cytokine production
protein dephosphorylation
actin filament organization
mitotic cell cycle checkpoint
establishment or maintenance of cell polarity
chemical synaptic transmission
nervous system development
positive regulation of cell proliferation
regulation of cell shape
viral process
single organismal cell-cell adhesion
peristalsis
positive regulation of actin filament polymerization
cortical actin cytoskeleton organization
astral microtubule organization
membrane raft organization
regulation of myelination
activation of protein kinase activity
ion transmembrane transport
cellular protein complex localization
T cell activation
negative regulation of T cell proliferation
regulation of membrane potential
amyloid precursor protein metabolic process
receptor clustering
positive regulation of potassium ion transport
cortical microtubule organization
establishment or maintenance of epithelial cell apical/basal polarity
negative regulation of mitotic cell cycle
GMP metabolic process
GDP metabolic process
reproductive structure development
embryonic skeletal system morphogenesis
smooth muscle tissue development
negative regulation of epithelial cell proliferation
establishment of centrosome localization
negative regulation of protein kinase B signaling
hard palate development
negative regulation of ERK1 and ERK2 cascade
bicellular tight junction assembly
protein localization to plasma membrane
positive regulation of establishment of protein localization to plasma membrane
receptor localization to synapse
regulation of NIK/NF-kappaB signaling
regulation of sodium ion transmembrane transport
negative regulation of p38MAPK cascade
guanylate kinase activity
phosphoprotein phosphatase activity
protein binding
protein C-terminus binding
cytoskeletal protein binding
ligand-gated ion channel activity
potassium channel regulator activity
protein kinase binding
phosphatase binding
mitogen-activated protein kinase kinase binding
protein complex scaffold
ionotropic glutamate receptor binding
ion channel binding
L27 domain binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0