Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNAP25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322902.1 | NP_001309831.1 | 5 - 6 | 550 | 133 | |
NM_003081.4 | NP_003072.2 | 5 - 6 | 519 | 133 | ||
XM_017028021.1 | XP_016883510.1 | 6 - 7 | 449 | 133 | ||
XM_017028022.1 | XP_016883511.1 | 6 - 7 | 541 | 133 | ||
GenBank mRNA | AK098767.1 | - | 5 - 6 | 389 | 133 | |
AK289647.1 | - | 7 - 8 | 680 | 133 | ||
BC010647.1 | - | 5 - 6 | 408 | 3867544 | 133 | |
BT019684.1 | - | 4 - 5 | 276 | 133 | ||
L19760.1 | - | 5 - 6 | 364 | 133 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |