Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTDSP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005730.3 | NP_005721.3 | 4 - 5 | 880 | 80 | |
XM_005268556.1 | XP_005268613.1 | 4 - 5 | 658 | 80 | ||
GenBank mRNA | AF000152.1 | - | 4 - 5 | 656 | 80 | |
AF022231.1 | - | 4 - 5 | 597 | 80 | ||
AK226076.1 | - | 4 - 5 | 636 | 80 | ||
AK291289.1 | - | 4 - 5 | 627 | 80 | ||
AK310873.1 | - | 4 - 5 | 382 | 80 | ||
AK312413.1 | - | 4 - 5 | 639 | 80 | ||
AY279531.1 | - | 4 - 5 | 351 | 80 | ||
BC018922.2 | - | 4 - 5 | 612 | 80 | ||
BC065920.1 | - | 4 - 5 | 595 | 5756196 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |