Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007533.hg.1 |
ENST00000340553.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007533.hg.1 |
ENST00000340553.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007533.hg.1 |
ENST00000340553.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007533.hg.1 |
ENST00000340553.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007533.hg.1 |
ENST00000340553.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007533.hg.1 |
ENST00000340553.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007533.hg.1 |
ENST00000340553.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007533.hg.1 |
ENST00000340553.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007533.hg.1 |
ENST00000340553.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007533.hg.1 |
ENST00000340553.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007533.hg.1 |
ENST00000340553.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007533.hg.1 |
ENST00000340553.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007533.hg.1 |
ENST00000413569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007533.hg.1 |
ENST00000413569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007533.hg.1 |
ENST00000413569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007533.hg.1 |
ENST00000413569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007533.hg.1 |
ENST00000413569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007533.hg.1 |
ENST00000413569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007533.hg.1 |
ENST00000616844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007533.hg.1 |
ENST00000616844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007533.hg.1 |
ENST00000616844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007533.hg.1 |
ENST00000616844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007533.hg.1 |
ENST00000616844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007533.hg.1 |
ENST00000616844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007533.hg.1 |
ENST00000616844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007533.hg.1 |
ENST00000616844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007533.hg.1 |
NM_002158.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007533.hg.1 |
NM_002158.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007533.hg.1 |
NM_002158.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007533.hg.1 |
NM_002158.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007533.hg.1 |
NM_002158.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007533.hg.1 |
NM_002158.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007533.hg.1 |
NM_002158.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007533.hg.1 |
NM_002158.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007533.hg.1 |
NM_002158.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007533.hg.1 |
NM_002158.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007533.hg.1 |
NM_002158.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007533.hg.1 |
NM_002158.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007533.hg.1 |
NM_002158.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004598.hg.1 |
TC02000302.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004598.hg.1 |
ENST00000304367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004598.hg.1 |
ENST00000340553.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004598.hg.1 |
NM_002158.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004597.hg.1 |
TC02000302.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004597.hg.1 |
ENST00000304367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004597.hg.1 |
ENST00000340553.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004597.hg.1 |
NM_002158.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200152117.hg.1 |
ENST00000340553.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200152117.hg.1 |
ENST00000413569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200152117.hg.1 |
ENST00000616844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200152117.hg.1 |
NM_002158.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200152117.hg.1 |
ENST00000340553.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200152117.hg.1 |
ENST00000413569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200152117.hg.1 |
ENST00000616844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200152117.hg.1 |
NM_002158.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200152117.hg.1 |
TC0200007533.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200152118.hg.1 |
ENST00000340553.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200152118.hg.1 |
ENST00000616844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200152118.hg.1 |
NM_002158.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200152118.hg.1 |
ENST00000340553.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200152118.hg.1 |
ENST00000616844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200152118.hg.1 |
NM_002158.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200152118.hg.1 |
TC0200007533.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200007533.hg.1 |
ENST00000340553.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007533.hg.1 |
ENST00000340553.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007533.hg.1 |
ENST00000340553.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007533.hg.1 |
ENST00000340553.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007533.hg.1 |
ENST00000340553.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007533.hg.1 |
ENST00000340553.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007533.hg.1 |
ENST00000340553.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007533.hg.1 |
ENST00000340553.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007533.hg.1 |
ENST00000340553.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007533.hg.1 |
ENST00000340553.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007533.hg.1 |
ENST00000340553.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007533.hg.1 |
ENST00000340553.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007533.hg.1 |
ENST00000413569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007533.hg.1 |
ENST00000413569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007533.hg.1 |
ENST00000413569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007533.hg.1 |
ENST00000413569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007533.hg.1 |
ENST00000413569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007533.hg.1 |
ENST00000413569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007533.hg.1 |
ENST00000616844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007533.hg.1 |
ENST00000616844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007533.hg.1 |
ENST00000616844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007533.hg.1 |
ENST00000616844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007533.hg.1 |
ENST00000616844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007533.hg.1 |
ENST00000616844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007533.hg.1 |
ENST00000616844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007533.hg.1 |
ENST00000616844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007533.hg.1 |
NM_002158.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007533.hg.1 |
NM_002158.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007533.hg.1 |
NM_002158.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007533.hg.1 |
NM_002158.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007533.hg.1 |
NM_002158.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007533.hg.1 |
NM_002158.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007533.hg.1 |
NM_002158.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007533.hg.1 |
NM_002158.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007533.hg.1 |
NM_002158.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007533.hg.1 |
NM_002158.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007533.hg.1 |
NM_002158.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007533.hg.1 |
NM_002158.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007533.hg.1 |
NM_002158.3 |
Refseq |