Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128431.2 | NP_001121903.1 | 4 - 5 | 803 | 83 | |
NM_001135153.1 | NP_001128625.1 | 4 - 5 | 844 | 83 | ||
NM_001135154.1 | NP_001128626.1 | 4 - 5 | 803 | 83 | ||
XM_005273465.2 | XP_005273522.1 | 5 - 6 | 891 | 83 | ||
XM_005273466.4 | XP_005273523.1 | 4 - 5 | 875 | 83 | ||
XM_006716323.2 | XP_006716386.1 | 6 - 7 | 1089 | 83 | ||
XM_006716324.2 | XP_006716387.1 | 5 - 6 | 880 | 83 | ||
XM_011544478.2 | XP_011542780.1 | 5 - 6 | 955 | 83 | ||
XM_017013292.1 | XP_016868781.1 | 5 - 6 | 971 | 83 | ||
XM_017013294.1 | XP_016868783.1 | 6 - 7 | 998 | 83 | ||
XM_017013295.1 | XP_016868784.1 | 4 - 5 | 1008 | 83 | ||
GenBank mRNA | AK295807.1 | - | 4 - 5 | 719 | 83 | |
BC015770.1 | - | 4 - 5 | 765 | 4865469 | 83 | |
D31887.1 | - | 4 - 5 | 747 | 83 | ||
KU178568.1 | - | 3 - 4 | 628 | 83 | ||
KU178569.1 | - | 3 - 4 | 628 | 83 | ||
KU178571.1 | - | 3 - 4 | 628 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons |