Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006816.hg.1 |
TC0X000511.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006816.hg.1 |
ENST00000344799.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006816.hg.1 |
ENST00000372582.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006816.hg.1 |
NM_017416.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008031.hg.1 |
ENST00000344799.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008031.hg.1 |
ENST00000344799.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008031.hg.1 |
ENST00000344799.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008031.hg.1 |
ENST00000344799.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008031.hg.1 |
ENST00000344799.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008031.hg.1 |
ENST00000344799.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008031.hg.1 |
ENST00000372582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008031.hg.1 |
ENST00000372582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008031.hg.1 |
ENST00000372582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008031.hg.1 |
ENST00000372582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008031.hg.1 |
ENST00000372582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008031.hg.1 |
ENST00000372582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008031.hg.1 |
ENST00000372582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008031.hg.1 |
ENST00000372582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008031.hg.1 |
ENST00000372582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008031.hg.1 |
ENST00000372582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008031.hg.1 |
ENST00000372582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008031.hg.1 |
ENST00000372582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008031.hg.1 |
ENST00000372582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008031.hg.1 |
ENST00000372582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008031.hg.1 |
ENST00000372582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008031.hg.1 |
ENST00000485671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008031.hg.1 |
ENST00000485671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008031.hg.1 |
ENST00000485671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008031.hg.1 |
ENST00000485671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008031.hg.1 |
NM_017416.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008031.hg.1 |
NM_017416.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008031.hg.1 |
NM_017416.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008031.hg.1 |
NM_017416.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008031.hg.1 |
NM_017416.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008031.hg.1 |
NM_017416.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008031.hg.1 |
NM_017416.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008031.hg.1 |
NM_017416.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008031.hg.1 |
NM_017416.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008031.hg.1 |
NM_017416.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008031.hg.1 |
NM_017416.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008031.hg.1 |
NM_017416.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008031.hg.1 |
NM_017416.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008031.hg.1 |
NM_017416.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008031.hg.1 |
NM_017416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00154903.hg.1 |
ENST00000372582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00154903.hg.1 |
NM_017416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00154903.hg.1 |
ENST00000372582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00154903.hg.1 |
NM_017416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00154903.hg.1 |
TC0X00008031.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00008031.hg.1 |
ENST00000344799.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008031.hg.1 |
ENST00000344799.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008031.hg.1 |
ENST00000344799.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008031.hg.1 |
ENST00000344799.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008031.hg.1 |
ENST00000344799.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008031.hg.1 |
ENST00000344799.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008031.hg.1 |
ENST00000372582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008031.hg.1 |
ENST00000372582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008031.hg.1 |
ENST00000372582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008031.hg.1 |
ENST00000372582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008031.hg.1 |
ENST00000372582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008031.hg.1 |
ENST00000372582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008031.hg.1 |
ENST00000372582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008031.hg.1 |
ENST00000372582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008031.hg.1 |
ENST00000372582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008031.hg.1 |
ENST00000372582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008031.hg.1 |
ENST00000372582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008031.hg.1 |
ENST00000372582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008031.hg.1 |
ENST00000372582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008031.hg.1 |
ENST00000372582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008031.hg.1 |
ENST00000372582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008031.hg.1 |
ENST00000485671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008031.hg.1 |
ENST00000485671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008031.hg.1 |
ENST00000485671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008031.hg.1 |
ENST00000485671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008031.hg.1 |
NM_017416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008031.hg.1 |
NM_017416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008031.hg.1 |
NM_017416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008031.hg.1 |
NM_017416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008031.hg.1 |
NM_017416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008031.hg.1 |
NM_017416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008031.hg.1 |
NM_017416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008031.hg.1 |
NM_017416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008031.hg.1 |
NM_017416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008031.hg.1 |
NM_017416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008031.hg.1 |
NM_017416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008031.hg.1 |
NM_017416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008031.hg.1 |
NM_017416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008031.hg.1 |
NM_017416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008031.hg.1 |
NM_017416.1 |
Refseq |