Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002111.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002111.hg.1 |
ENST00000398341.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002111.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002111.hg.1 |
TC21000202.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000398343.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000398343.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000398343.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000398343.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000398343.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000471277.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000471277.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000471277.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000471277.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000471277.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000484887.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000484887.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000484887.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000487951.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000487951.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000487951.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000496416.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000496416.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000496416.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000496416.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
ENST00000496416.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002113.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002113.hg.1 |
ENST00000398341.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002113.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002113.hg.1 |
TC21000202.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002112.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002112.hg.1 |
ENST00000398341.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002112.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002112.hg.1 |
TC21000202.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2100158351.hg.1 |
TC2100008527.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100158351.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158351.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158351.hg.1 |
ENST00000496416.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158351.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100158351.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158351.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158351.hg.1 |
ENST00000496416.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158351.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000398343.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000398343.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000398343.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000398343.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000398343.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000471277.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000471277.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000471277.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000471277.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000471277.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000484887.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000484887.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000484887.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000487951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000487951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000487951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000496416.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000496416.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000496416.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000496416.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
ENST00000496416.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008527.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100158352.hg.1 |
TC2100008527.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100158352.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158352.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158352.hg.1 |
ENST00000496416.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158352.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100158352.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158352.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158352.hg.1 |
ENST00000496416.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158352.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100158350.hg.1 |
TC2100008527.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100158350.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158350.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158350.hg.1 |
ENST00000496416.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158350.hg.1 |
NM_018964.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100158350.hg.1 |
ENST00000352133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158350.hg.1 |
ENST00000398341.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158350.hg.1 |
ENST00000496416.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158350.hg.1 |
NM_018964.3 |
Refseq |