Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013348.hg.1 |
ENST00000296642.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013348.hg.1 |
ENST00000296642.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013348.hg.1 |
ENST00000296642.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013348.hg.1 |
ENST00000296642.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013348.hg.1 |
ENST00000296642.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013348.hg.1 |
ENST00000296642.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013348.hg.1 |
ENST00000379615.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013348.hg.1 |
ENST00000379615.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013348.hg.1 |
ENST00000379615.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013348.hg.1 |
ENST00000379615.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013348.hg.1 |
ENST00000379615.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013348.hg.1 |
ENST00000379615.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013348.hg.1 |
ENST00000379615.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013348.hg.1 |
ENST00000379615.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013348.hg.1 |
ENST00000503638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013348.hg.1 |
ENST00000503638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013348.hg.1 |
ENST00000503638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013348.hg.1 |
ENST00000503638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013348.hg.1 |
ENST00000514178.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013348.hg.1 |
ENST00000514178.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013348.hg.1 |
ENST00000514178.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013348.hg.1 |
ENST00000514178.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013348.hg.1 |
NM_005023.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013348.hg.1 |
NM_005023.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013348.hg.1 |
NM_005023.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013348.hg.1 |
NM_005023.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013348.hg.1 |
NM_005023.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013348.hg.1 |
NM_005023.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013348.hg.1 |
NM_005023.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013348.hg.1 |
NM_005023.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013348.hg.1 |
NM_005023.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013348.hg.1 |
NM_005023.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013348.hg.1 |
NM_005023.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022883.hg.1 |
TC05001687.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022883.hg.1 |
ENST00000379615.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022883.hg.1 |
ENST00000419445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022883.hg.1 |
NM_005023.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013348.hg.1 |
ENST00000296642.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013348.hg.1 |
ENST00000296642.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013348.hg.1 |
ENST00000296642.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013348.hg.1 |
ENST00000296642.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013348.hg.1 |
ENST00000296642.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013348.hg.1 |
ENST00000296642.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013348.hg.1 |
ENST00000379615.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013348.hg.1 |
ENST00000379615.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013348.hg.1 |
ENST00000379615.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013348.hg.1 |
ENST00000379615.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013348.hg.1 |
ENST00000379615.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013348.hg.1 |
ENST00000379615.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013348.hg.1 |
ENST00000379615.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013348.hg.1 |
ENST00000379615.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013348.hg.1 |
ENST00000503638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013348.hg.1 |
ENST00000503638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013348.hg.1 |
ENST00000503638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013348.hg.1 |
ENST00000503638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013348.hg.1 |
ENST00000514178.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013348.hg.1 |
ENST00000514178.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013348.hg.1 |
ENST00000514178.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013348.hg.1 |
ENST00000514178.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013348.hg.1 |
NM_005023.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013348.hg.1 |
NM_005023.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013348.hg.1 |
NM_005023.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013348.hg.1 |
NM_005023.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013348.hg.1 |
NM_005023.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013348.hg.1 |
NM_005023.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013348.hg.1 |
NM_005023.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013348.hg.1 |
NM_005023.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013348.hg.1 |
NM_005023.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013348.hg.1 |
NM_005023.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013348.hg.1 |
NM_005023.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500196186.hg.1 |
ENST00000296642.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500196186.hg.1 |
ENST00000379615.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500196186.hg.1 |
NM_005023.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500196186.hg.1 |
ENST00000296642.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500196186.hg.1 |
ENST00000379615.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500196186.hg.1 |
NM_005023.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500196186.hg.1 |
TC0500013348.hg.1 |
Clariom D TC |