Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000328249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000328249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000328249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000328249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000328249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000328249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000328249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000328249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000328249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000328249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000328249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000328249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000372514.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000372514.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000372514.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000372514.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000372514.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000372514.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000372514.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000372514.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000372514.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000372514.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000372514.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000372514.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000372517.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000372517.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000372517.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000372517.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000372517.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000372517.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000372517.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000372517.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000372517.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000372517.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000372517.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000372517.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000470938.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000470938.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000470938.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000487556.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000487556.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000487556.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000487556.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000487556.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000487556.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000487556.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000487556.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000487556.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
ENST00000487556.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
NM_001017922.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
NM_001017922.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
NM_001017922.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
NM_001017922.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
NM_001017922.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
NM_001017922.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
NM_001017922.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
NM_001017922.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
NM_001017922.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
NM_001017922.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
NM_001017922.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
NM_001017922.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
NM_018538.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
NM_018538.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
NM_018538.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
NM_018538.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
NM_018538.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
NM_018538.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
NM_018538.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
NM_018538.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
NM_018538.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
NM_018538.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
NM_018538.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008033.hg.1 |
NM_018538.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01008452.hg.1 |
TC01000534.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01008452.hg.1 |
ENST00000328249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01008452.hg.1 |
ENST00000372514.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01008452.hg.1 |
ENST00000372517.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01008452.hg.1 |
ENST00000487556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01008452.hg.1 |
NM_001017922.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01008452.hg.1 |
NM_018538.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100156660.hg.1 |
ENST00000328249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100156660.hg.1 |
ENST00000372514.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100156660.hg.1 |
ENST00000372517.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100156660.hg.1 |
ENST00000487556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100156660.hg.1 |
NM_001017922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100156660.hg.1 |
NM_018538.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100156660.hg.1 |
ENST00000328249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100156660.hg.1 |
ENST00000372514.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100156660.hg.1 |
ENST00000372517.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100156660.hg.1 |
ENST00000487556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100156660.hg.1 |
NM_001017922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100156660.hg.1 |
NM_018538.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100156660.hg.1 |
TC0100008033.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000328249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000328249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000328249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000328249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000328249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000328249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000328249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000328249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000328249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000328249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000328249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000328249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000372514.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000372514.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000372514.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000372514.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000372514.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000372514.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000372514.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000372514.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000372514.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000372514.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000372514.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000372514.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000372517.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000372517.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000372517.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000372517.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000372517.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000372517.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000372517.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000372517.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000372517.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000372517.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000372517.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000372517.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000470938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000470938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000470938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000487556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000487556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000487556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000487556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000487556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000487556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000487556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000487556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000487556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
ENST00000487556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
NM_001017922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
NM_001017922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
NM_001017922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
NM_001017922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
NM_001017922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
NM_001017922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
NM_001017922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
NM_001017922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
NM_001017922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
NM_001017922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
NM_001017922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
NM_001017922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
NM_018538.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
NM_018538.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
NM_018538.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
NM_018538.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
NM_018538.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
NM_018538.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
NM_018538.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
NM_018538.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
NM_018538.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
NM_018538.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
NM_018538.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008033.hg.1 |
NM_018538.3 |
Refseq |