Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
ENST00000354618.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
ENST00000354618.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
ENST00000354618.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
ENST00000354618.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
ENST00000354618.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
ENST00000354618.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
ENST00000354618.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
ENST00000354618.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
ENST00000354618.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
ENST00000354618.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
ENST00000354618.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
ENST00000354618.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
ENST00000354618.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
ENST00000354618.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
ENST00000354618.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
ENST00000354618.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
ENST00000466743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
ENST00000466743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
ENST00000466743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
ENST00000466743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
ENST00000466743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
ENST00000469141.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
ENST00000469141.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
ENST00000469141.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
ENST00000469141.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
ENST00000469141.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
ENST00000469141.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
ENST00000469141.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
ENST00000469141.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
ENST00000469141.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
ENST00000469141.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
ENST00000469141.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
ENST00000469141.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
ENST00000469141.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
ENST00000469141.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
ENST00000469141.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
ENST00000469141.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
ENST00000478853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
ENST00000478853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
ENST00000478853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
ENST00000478853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
ENST00000478853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
NM_005119.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
NM_005119.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
NM_005119.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
NM_005119.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
NM_005119.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
NM_005119.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
NM_005119.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
NM_005119.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
NM_005119.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
NM_005119.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
NM_005119.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
NM_005119.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
NM_005119.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
NM_005119.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
NM_005119.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007798.hg.1 |
NM_005119.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006940.hg.1 |
ENST00000411753.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006940.hg.1 |
ENST00000411753.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006940.hg.1 |
ENST00000411753.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006940.hg.1 |
ENST00000411753.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006940.hg.1 |
ENST00000411753.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006940.hg.1 |
ENST00000411753.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01007284.hg.1 |
TC01000469.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01007284.hg.1 |
ENST00000354618.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01007284.hg.1 |
ENST00000469141.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01007284.hg.1 |
NM_005119.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100154977.hg.1 |
ENST00000354618.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154977.hg.1 |
ENST00000469141.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154977.hg.1 |
ENST00000478853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154977.hg.1 |
NM_005119.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100154977.hg.1 |
ENST00000354618.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154977.hg.1 |
ENST00000469141.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154977.hg.1 |
ENST00000478853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154977.hg.1 |
NM_005119.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100154977.hg.1 |
TC0100007798.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100154976.hg.1 |
ENST00000354618.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154976.hg.1 |
ENST00000469141.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154976.hg.1 |
ENST00000478853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154976.hg.1 |
NM_005119.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100154976.hg.1 |
ENST00000354618.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154976.hg.1 |
ENST00000469141.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154976.hg.1 |
ENST00000478853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154976.hg.1 |
NM_005119.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100154976.hg.1 |
TC0100007798.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100154973.hg.1 |
ENST00000469141.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154973.hg.1 |
ENST00000469141.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154973.hg.1 |
TC0100007798.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
ENST00000354618.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
ENST00000354618.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
ENST00000354618.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
ENST00000354618.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
ENST00000354618.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
ENST00000354618.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
ENST00000354618.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
ENST00000354618.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
ENST00000354618.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
ENST00000354618.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
ENST00000354618.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
ENST00000354618.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
ENST00000354618.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
ENST00000354618.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
ENST00000354618.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
ENST00000354618.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
ENST00000466743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
ENST00000466743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
ENST00000466743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
ENST00000466743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
ENST00000466743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
ENST00000469141.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
ENST00000469141.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
ENST00000469141.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
ENST00000469141.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
ENST00000469141.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
ENST00000469141.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
ENST00000469141.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
ENST00000469141.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
ENST00000469141.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
ENST00000469141.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
ENST00000469141.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
ENST00000469141.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
ENST00000469141.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
ENST00000469141.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
ENST00000469141.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
ENST00000469141.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
ENST00000478853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
ENST00000478853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
ENST00000478853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
ENST00000478853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
ENST00000478853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
NM_005119.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
NM_005119.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
NM_005119.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
NM_005119.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
NM_005119.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
NM_005119.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
NM_005119.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
NM_005119.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
NM_005119.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
NM_005119.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
NM_005119.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
NM_005119.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
NM_005119.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
NM_005119.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
NM_005119.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007798.hg.1 |
NM_005119.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006940.hg.1 |
ENST00000411753.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006940.hg.1 |
ENST00000411753.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006940.hg.1 |
ENST00000411753.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006940.hg.1 |
ENST00000411753.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006940.hg.1 |
ENST00000411753.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006940.hg.1 |
ENST00000411753.1 |
Ensembl |