Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PATE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138294.2 | NP_612151.1 | 3 - 4 | 136 | 117 | |
GenBank mRNA | AF462605.1 | - | 3 - 4 | 132 | 117 | |
BC039863.1 | - | 3 - 4 | 136 | 5745088 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |