Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028056.hg.1 |
TC06002238.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028056.hg.1 |
ENST00000265368.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028056.hg.1 |
ENST00000341594.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028056.hg.1 |
ENST00000367255.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028056.hg.1 |
ENST00000423061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028056.hg.1 |
ENST00000448038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028056.hg.1 |
NM_033071.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028056.hg.1 |
NM_182961.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193906.hg.1 |
ENST00000341594.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193906.hg.1 |
ENST00000367255.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193906.hg.1 |
ENST00000423061.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193906.hg.1 |
ENST00000461872.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193906.hg.1 |
NM_033071.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193906.hg.1 |
NM_182961.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193906.hg.1 |
ENST00000341594.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193906.hg.1 |
ENST00000367255.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193906.hg.1 |
ENST00000423061.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193906.hg.1 |
ENST00000461872.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193906.hg.1 |
NM_033071.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193906.hg.1 |
NM_182961.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193906.hg.1 |
TC0600013588.hg.1 |
Clariom D TC |