Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
ENST00000377067.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
ENST00000377067.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
ENST00000377067.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
ENST00000377067.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
ENST00000377067.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
ENST00000377067.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
ENST00000377067.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
ENST00000377067.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
ENST00000377067.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
ENST00000377067.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
ENST00000483422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
ENST00000483422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
ENST00000483422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
ENST00000618283.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
ENST00000618283.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
ENST00000618596.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
ENST00000618596.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
ENST00000618596.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003413.hg.1 |
ENST00000377067.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003413.hg.1 |
NM_004466.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003413.hg.1 |
TC13000323.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
ENST00000377067.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
ENST00000377067.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
ENST00000377067.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
ENST00000377067.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
ENST00000377067.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
ENST00000377067.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
ENST00000377067.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
ENST00000377067.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
ENST00000377067.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
ENST00000377067.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
ENST00000483422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
ENST00000483422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
ENST00000483422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
ENST00000618283.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
ENST00000618283.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
ENST00000618596.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
ENST00000618596.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
ENST00000618596.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152052.hg.1 |
ENST00000377067.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152052.hg.1 |
ENST00000618596.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152052.hg.1 |
NM_004466.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152052.hg.1 |
ENST00000377067.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152052.hg.1 |
ENST00000618596.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152052.hg.1 |
NM_004466.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152052.hg.1 |
TC1300007707.hg.1 |
Clariom D TC |