Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023646.hg.1 |
TC06001929.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023646.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023646.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023645.hg.1 |
TC06001929.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023645.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023645.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023648.hg.1 |
TC06001929.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023648.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023648.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023647.hg.1 |
TC06001929.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023647.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023647.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008967.hg.1 |
ENST00000505898.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023650.hg.1 |
TC06001929.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023650.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
ENST00000420494.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
ENST00000420494.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186212.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186212.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186212.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186212.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186212.hg.1 |
TC0600012514.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600186222.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186222.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186222.hg.1 |
TC0600012514.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600186219.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186219.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186219.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186219.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186219.hg.1 |
TC0600012514.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
ENST00000420494.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
ENST00000420494.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186213.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186213.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186213.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186213.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186213.hg.1 |
TC0600012514.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600186217.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186217.hg.1 |
ENST00000420494.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186217.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186217.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186217.hg.1 |
ENST00000420494.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186217.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186217.hg.1 |
TC0600012514.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600186218.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186218.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186218.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186218.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186218.hg.1 |
TC0600012514.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600186220.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186220.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186220.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186220.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186220.hg.1 |
TC0600012514.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400008967.hg.1 |
ENST00000505898.1 |
Ensembl |