Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
ENST00000217026.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
ENST00000217026.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
ENST00000217026.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
ENST00000217026.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
ENST00000217026.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
ENST00000217026.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
ENST00000217026.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
ENST00000217026.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
ENST00000217026.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
ENST00000217026.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
ENST00000217026.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
ENST00000217026.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
ENST00000217026.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
ENST00000217026.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
ENST00000217026.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
ENST00000396863.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
ENST00000396863.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
ENST00000396863.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
ENST00000396863.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
ENST00000396863.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
ENST00000396863.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
ENST00000396863.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
ENST00000396863.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
ENST00000396863.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
ENST00000396863.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
ENST00000396863.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
ENST00000396863.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
ENST00000396863.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
ENST00000396863.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
ENST00000396863.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
ENST00000396863.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
ENST00000396863.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
ENST00000396863.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
NM_001278610.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
NM_001278610.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
NM_001278610.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
NM_001278610.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
NM_001278610.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
NM_001278610.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
NM_001278610.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
NM_001278610.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
NM_001278610.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
NM_001278610.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
NM_001278610.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
NM_001278610.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
NM_001278610.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
NM_001278610.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
NM_001278610.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
NM_001278610.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
NM_001278610.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
NM_001278610.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
NM_002466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
NM_002466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
NM_002466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
NM_002466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
NM_002466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
NM_002466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
NM_002466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
NM_002466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
NM_002466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
NM_002466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
NM_002466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
NM_002466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
NM_002466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
NM_002466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
NM_002466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
NM_002466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
NM_002466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
NM_002466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007438.hg.1 |
NM_002466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000151531.hg.1 |
ENST00000217026.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000151531.hg.1 |
ENST00000396863.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000151531.hg.1 |
NM_001278610.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000151531.hg.1 |
NM_002466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000151531.hg.1 |
TC2000007438.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000151531.hg.1 |
ENST00000217026.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000151531.hg.1 |
ENST00000396863.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000151531.hg.1 |
NM_001278610.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000151531.hg.1 |
NM_002466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
ENST00000217026.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
ENST00000217026.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
ENST00000217026.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
ENST00000217026.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
ENST00000217026.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
ENST00000217026.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
ENST00000217026.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
ENST00000217026.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
ENST00000217026.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
ENST00000217026.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
ENST00000217026.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
ENST00000217026.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
ENST00000217026.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
ENST00000217026.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
ENST00000217026.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
ENST00000396863.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
ENST00000396863.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
ENST00000396863.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
ENST00000396863.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
ENST00000396863.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
ENST00000396863.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
ENST00000396863.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
ENST00000396863.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
ENST00000396863.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
ENST00000396863.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
ENST00000396863.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
ENST00000396863.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
ENST00000396863.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
ENST00000396863.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
ENST00000396863.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
ENST00000396863.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
ENST00000396863.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
ENST00000396863.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
NM_001278610.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
NM_001278610.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
NM_001278610.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
NM_001278610.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
NM_001278610.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
NM_001278610.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
NM_001278610.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
NM_001278610.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
NM_001278610.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
NM_001278610.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
NM_001278610.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
NM_001278610.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
NM_001278610.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
NM_001278610.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
NM_001278610.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
NM_001278610.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
NM_001278610.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
NM_001278610.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
NM_002466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
NM_002466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
NM_002466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
NM_002466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
NM_002466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
NM_002466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
NM_002466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
NM_002466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
NM_002466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
NM_002466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
NM_002466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
NM_002466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
NM_002466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
NM_002466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
NM_002466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
NM_002466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
NM_002466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
NM_002466.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007438.hg.1 |
NM_002466.3 |
Refseq |