Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011642.hg.1 |
ENST00000259938.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011642.hg.1 |
ENST00000259938.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011642.hg.1 |
ENST00000259938.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011642.hg.1 |
ENST00000259938.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011642.hg.1 |
ENST00000259938.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011642.hg.1 |
ENST00000616014.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011642.hg.1 |
ENST00000616014.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011642.hg.1 |
ENST00000616014.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011642.hg.1 |
ENST00000616014.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011642.hg.1 |
ENST00000622413.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011642.hg.1 |
ENST00000622413.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011642.hg.1 |
ENST00000622413.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011642.hg.1 |
ENST00000622413.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011642.hg.1 |
ENST00000622413.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011642.hg.1 |
NM_001252597.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011642.hg.1 |
NM_001252597.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011642.hg.1 |
NM_001252597.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011642.hg.1 |
NM_001252597.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011642.hg.1 |
NM_001252597.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011642.hg.1 |
NM_001252597.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011642.hg.1 |
NM_001252597.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011642.hg.1 |
NM_001252598.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011642.hg.1 |
NM_001252598.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011642.hg.1 |
NM_001252598.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011642.hg.1 |
NM_001252598.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011642.hg.1 |
NM_001832.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011642.hg.1 |
NM_001832.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011642.hg.1 |
NM_001832.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011642.hg.1 |
NM_001832.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011642.hg.1 |
NM_001832.3 |
Refseq |